Product References
Matrix-bound nanovesicle-associated IL-33 supports functional recovery after skeletal muscle injury by initiating a pro-regenerative macrophage phenotypic transition.
NPJ Regenerative medicine
Bartolacci JG,Behun MN,Warunek JP,Li T,Sahu A,Dwyer GK,Lucas A,Rong J,Ambrosio F,Turnquist HR,Badylak SF
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Jan 27 00:00:00 UTC 2024
High-Dose Acetaminophen with N-acetylcysteine Rescue Inhibits M2 Polarization of Tumor-Associated Macrophages.
Cancers
Bryan A,Pingali P,Joslyn M,Li H,Bernas T,Koblinski J,Landry J,Lee WS,Patel B,Neuwelt A
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Sep 28 00:00:00 UTC 2023
Janus porous polylactic acid membranes with versatile metal-phenolic interface for biomimetic periodontal bone regeneration.
NPJ Regenerative medicine
Zhang Y,Chen Y,Ding T,Zhang Y,Yang D,Zhao Y,Liu J,Ma B,Bianco A,Ge S,Li J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Jun 03 00:00:00 UTC 2023
Acute murine cytomegalovirus infection boosts cell-type specific response and lipid metabolism changes in the liver of infant mice.
Frontiers in immunology
Gao J,Wang A,Bu X,Jia W
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Aug 29 00:00:00 UTC 2023
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2023
SARS-CoV-2 N protein induced acute kidney injury in diabetic db/db mice is associated with a Mincle-dependent M1 macrophage activation.
Frontiers in immunology
Wu W,Wang W,Liang L,Chen J,Sun S,Wei B,Zhong Y,Huang XR,Liu J,Wang X,Yu X,Lan HY
17-5920-82 was used in Flow Cytometry to find that quercetin treatment significantly inhibited the release of a DAMP molecule HMGB1 and inactivated M1 pro-inflammatory macrophage while promoting reparative M2 macrophage responses by suppressing Mincle-Syk/NF-κB signaling in vivo and in vitro.
Fri Dec 01 00:00:00 UTC 2023
A druggable copper-signalling pathway that drives inflammation.
Nature
Solier S,Müller S,Cañeque T,Versini A,Mansart A,Sindikubwabo F,Baron L,Emam L,Gestraud P,Pantoș GD,Gandon V,Gaillet C,Wu TD,Dingli F,Loew D,Baulande S,Durand S,Sencio V,Robil C,Trottein F,Péricat D,Näser E,Cougoule C,Meunier E,Bègue AL,Salmon H,Manel N,Puisieux A,Watson S,Dawson MA,Servant N,Kroemer G,Annane D,Rodriguez R
17-5920-82 was used in Flow Cytometry to highlight the central role of copper as a regulator of cell plasticity and unveils a therapeutic strategy based on metabolic reprogramming and the control of epigenetic cell states.
Mon May 01 00:00:00 UTC 2023
Functionally and Metabolically Divergent Melanoma-Associated Macrophages Originate from Common Bone-Marrow Precursors.
Cancers
Pizzurro GA,Bridges K,Jiang X,Vidyarthi A,Miller-Jensen K,Colegio OR
17-5920-82 was used in Flow Cytometry to study melanoma Tumor-associated macrophages (TAMs) origin and dynamics during tumor progression.
Sat Jun 24 00:00:00 UTC 2023
IL-18BP mediates the balance between protective and pathological immune responses to Toxoplasma gondii.
Cell reports
Clark JT,Weizman OE,Aldridge DL,Shallberg LA,Eberhard J,Lanzar Z,Wasche D,Huck JD,Zhou T,Ring AM,Hunter CA
17-5920-82 was used in Flow cytometry/Cell sorting to investigate the role of interleukin-18 (IL-18) and its binding protein (IL-18BP) in controlling Toxoplasma gondii infection.
Tue Mar 28 00:00:00 UTC 2023
ROS-responsive polymer nanoparticles with enhanced loading of dexamethasone effectively modulate the lung injury microenvironment.
Acta biomaterialia
Muhammad W,Zhu J,Zhai Z,Xie J,Zhou J,Feng X,Feng B,Pan Q,Li S,Venkatesan R,Li P,Cao H,Gao C
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
EGFR tyrosine kinase activity and Rab GTPases coordinate EGFR trafficking to regulate macrophage activation in sepsis.
Cell death & disease
Zhang X,Chen C,Ling C,Luo S,Xiong Z,Liu X,Liao C,Xie P,Liu Y,Zhang L,Chen Z,Liu Z,Tang J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Nov 07 00:00:00 UTC 2022
Cutting Edge: Enhanced Antitumor Immunity in ST8Sia6 Knockout Mice.
Journal of immunology (Baltimore, Md. : 1950)
Friedman DJ,Kizerwetter M,Belmonte P,Rajcula M,Theodore K,Kim Lee HS,Shapiro MJ,Dong H,Shapiro VS
17-5920-82 was used in Flow cytometry/Cell sorting to examine the role of ST8Sia6 on immune cells in regulating antitumor immunity. ST8Sia6 knockout mice had an enhanced immune response to tumors.
Fri Apr 15 00:00:00 UTC 2022
Neuropeptide Y attenuates cardiac remodeling and deterioration of function following myocardial infarction.
Molecular therapy : the journal of the American Society of Gene Therapy
Qin YY,Huang XR,Zhang J,Wu W,Chen J,Wan S,Yu XY,Lan HY
17-5920-82 was used in Flow cytometry/Cell sorting to find that AMI mice lacking NPY developed more severe AMI than WT mice with worse cardiac dysfunction, progressive cardiac inflammation and fibrosis, and excessive apoptosis but impairing angiogenesis.
Wed Feb 02 00:00:00 UTC 2022
Nitric oxide controls proliferation of Leishmania major by inhibiting the recruitment of permissive host cells.
Immunity
Formaglio P,Alabdullah M,Siokis A,Handschuh J,Sauerland I,Fu Y,Krone A,Gintschel P,Stettin J,Heyde S,Mohr J,Philipsen L,Schröder A,Robert PA,Zhao G,Khailaie S,Dudeck A,Bertrand J,Späth GF,Kahlfuß S,Bousso P,Schraven B,Huehn J,Binder S,Meyer-Hermann M,Müller AJ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Dec 14 00:00:00 UTC 2021
Nitric oxide controls proliferation of Leishmania major by inhibiting the recruitment of permissive host cells.
Immunity
Formaglio P,Alabdullah M,Siokis A,Handschuh J,Sauerland I,Fu Y,Krone A,Gintschel P,Stettin J,Heyde S,Mohr J,Philipsen L,Schröder A,Robert PA,Zhao G,Khailaie S,Dudeck A,Bertrand J,Späth GF,Kahlfuß S,Bousso P,Schraven B,Huehn J,Binder S,Meyer-Hermann M,Müller AJ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Dec 14 00:00:00 UTC 2021
Immunogenic Chemotherapy Enhances Recruitment of CAR-T Cells to Lung Tumors and Improves Antitumor Efficacy when Combined with Checkpoint Blockade.
Cancer cell
Srivastava S,Furlan SN,Jaeger-Ruckstuhl CA,Sarvothama M,Berger C,Smythe KS,Garrison SM,Specht JM,Lee SM,Amezquita RA,Voillet V,Muhunthan V,Yechan-Gunja S,Pillai SPS,Rader C,Houghton AM,Pierce RH,Gottardo R,Maloney DG,Riddell SR
17-5920-82 was used in Flow Cytometry to test strategies for enhancing efficacy of CAR-T cells targeting the tumor-associated antigen receptor tyrosine kinase-like orphan receptor 1 (ROR1) infiltrating tumors.
Mon Feb 08 00:00:00 UTC 2021
Neutrophil-Macrophage Imbalance Drives the Development of Renal Scarring during Experimental Pyelonephritis.
Journal of the American Society of Nephrology : JASN
Ruiz-Rosado JD,Robledo-Avila F,Cortado H,Rangel-Moreno J,Justice SS,Yang C,Spencer JD,Becknell B,Partida-Sanchez S
17-5920-82 was used in Flow Cytometry to reveal a previously unappreciated outcome for neutrophil-macrophage imbalance in promoting host susceptibility to acute pyelonephritis and the development of permanent renal damage.
Fri Jan 01 00:00:00 UTC 2021
AXL Is a Key Factor for Cell Plasticity and Promotes Metastasis in Pancreatic Cancer.
Molecular cancer research : MCR
Du W,Phinney NZ,Huang H,Wang Z,Westcott J,Toombs JE,Zhang Y,Beg MS,Wilkie TM,Lorens JB,Brekken RA
17-5920-82 was used in Western Blot to implicate AXL as a marker of undifferentiated PDA cells and a target for therapy.
Sun Aug 01 00:00:00 UTC 2021
Prostaglandin E3 attenuates macrophage-associated inflammation and prostate tumour growth by modulating polarization.
Journal of cellular and molecular medicine
Cui J,Shan K,Yang Q,Qi Y,Qu H,Li J,Wang R,Jia L,Chen W,Feng N,Chen YQ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Jun 01 00:00:00 UTC 2021
Regulation of the long noncoding RNA XIST on the inflammatory polarization of microglia in cerebral infarction.
Experimental and therapeutic medicine
Zhang M,Yang JK,Ma J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Sep 01 00:00:00 UTC 2021
Exosomes from human umbilical cord mesenchymal stem cells attenuate the inflammation of severe steroid-resistant asthma by reshaping macrophage polarization.
Stem cell research & therapy
Dong B,Wang C,Zhang J,Zhang J,Gu Y,Guo X,Zuo X,Pan H,Hsu AC,Wang G,Wang F
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Mar 24 00:00:00 UTC 2021
Alphavirus-Driven Interferon Gamma (IFNg) Expression Inhibits Tumor Growth in Orthotopic 4T1 Breast Cancer Model.
Vaccines
Trofimova O,Korotkaja K,Skrastina D,Jansons J,Spunde K,Isaguliants M,Zajakina A
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Oct 27 00:00:00 UTC 2021
PD-L1 Improves Motor Function and Alleviates Neuropathic Pain in Male Mice After Spinal Cord Injury by Inhibiting MAPK Pathway.
Frontiers in immunology
Kong F,Sun K,Zhu J,Li F,Lin F,Sun X,Luo X,Ren C,Lu L,Zhao S,Sun J,Wang Y,Shi J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Nov 01 00:00:00 UTC 2021
TREM2 is a receptor for non-glycosylated mycolic acids of mycobacteria that limits anti-mycobacterial macrophage activation.
Nature communications
Iizasa E,Chuma Y,Uematsu T,Kubota M,Kawaguchi H,Umemura M,Toyonaga K,Kiyohara H,Yano I,Colonna M,Sugita M,Matsuzaki G,Yamasaki S,Yoshida H,Hara H
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Apr 16 00:00:00 UTC 2021
Computational modeling reveals a key role for polarized myeloid cells in controlling osteoclast activity during bone injury repair.
Scientific reports
Lo CH,Baratchart E,Basanta D,Lynch CC
17-5920 was used in Flow cytometry/Cell sorting to underscore the value of mathematical modeling approaches to understand and reveal new insights into complex biological processes.
Mon Mar 15 00:00:00 UTC 2021
Computational modeling reveals a key role for polarized myeloid cells in controlling osteoclast activity during bone injury repair.
Scientific reports
Lo CH,Baratchart E,Basanta D,Lynch CC
17-5920 was used in Flow cytometry/Cell sorting to underscore the value of mathematical modeling approaches to understand and reveal new insights into complex biological processes.
Mon Mar 15 00:00:00 UTC 2021
3D Model of the Early Melanoma Microenvironment Captures Macrophage Transition into a Tumor-Promoting Phenotype.
Cancers
Pizzurro GA,Liu C,Bridges K,Alexander AF,Huang A,Baskaran JP,Ramseier J,Bosenberg MW,Mak M,Miller-Jensen K
17-5920-82 was used in Flow cytometry/Cell sorting to develop 3D in vitro co-culture systems, composed of a collagen-I matrix scaffolding murine bone-marrow-derived macrophages (BMDMs), YUMM1.7 melanoma cells, and fibroblasts to recreate the early melanoma TME and study how interactions with fibroblasts and tumor cells modulate macrophage immune activity.
Sun Sep 12 00:00:00 UTC 2021
Macrophages Expressing GALC Improve Peripheral Krabbe Disease by a Mechanism Independent of Cross-Correction.
Neuron
Weinstock NI,Shin D,Dhimal N,Hong X,Irons EE,Silvestri NJ,Reed CB,Nguyen D,Sampson O,Cheng YC,Lau JTY,Bongarzone ER,Kofler J,Escolar ML,Gelb MH,Wrabetz L,Feltri ML
17-5920 was used in Flow cytometry/Cell sorting to show that cross-correction does not occur efficiently in vivo and that Galc-deficient Schwann cells autonomously produce psychosine.
Wed Jul 08 00:00:00 UTC 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Astrocytes promote a protective immune response to brain Toxoplasma gondii infection via IL-33-ST2 signaling.
PLoS pathogens
Still KM,Batista SJ,O'Brien CA,Oyesola OO,Früh SP,Webb LM,Smirnov I,Kovacs MA,Cowan MN,Hayes NW,Thompson JA,Tait Wojno ED,Harris TH
17-5920 was used in Flow cytometry/Cell sorting to show that IL-33 is expressed by oligodendrocytes and astrocytes during T. gondii infection, is released locally into the cerebrospinal fluid of T. gondii-infected animals, and is required for control of infection.
Thu Oct 01 00:00:00 UTC 2020
Anticancer Activity of Liquid Treated with Microwave Plasma-Generated Gas through Macrophage Activation.
Oxidative medicine and cellular longevity
Lee CB,Seo IH,Chae MW,Park JW,Choi EH,Uhm HS,Baik KY
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Sep 17 00:00:00 UTC 2020
Mycobacterium tuberculosis-infected alveolar epithelial cells modulate dendritic cell function through the HIF-1α-NOS2 axis.
Journal of leukocyte biology
Rodrigues TS,Alvarez ARP,Gembre AF,Forni MFPAD,de Melo BMS,Alves Filho JCF,Câmara NOS,Bonato VLD
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Enhanced alleviation of aGVHD by TGF-β1-modified mesenchymal stem cells in mice through shifting MΦ into M2 phenotype and promoting the differentiation of Treg cells.
Journal of cellular and molecular medicine
Wu R,Liu C,Deng X,Chen L,Hao S,Ma L
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
Proimmunogenic impact of MEK inhibition synergizes with agonist anti-CD40 immunostimulatory antibodies in tumor therapy.
Nature communications
Baumann D,Hägele T,Mochayedi J,Drebant J,Vent C,Blobner S,Noll JH,Nickel I,Schumacher C,Boos SL,Daniel AS,Wendler S,Volkmar M,Strobel O,Offringa R
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Proimmunogenic impact of MEK inhibition synergizes with agonist anti-CD40 immunostimulatory antibodies in tumor therapy.
Nature communications
Baumann D,Hägele T,Mochayedi J,Drebant J,Vent C,Blobner S,Noll JH,Nickel I,Schumacher C,Boos SL,Daniel AS,Wendler S,Volkmar M,Strobel O,Offringa R
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
CCL2 and CXCL12 Derived from Mesenchymal Stromal Cells Cooperatively Polarize IL-10+ Tissue Macrophages to Mitigate Gut Injury.
Cell reports
Giri J,Das R,Nylen E,Chinnadurai R,Galipeau J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Feb 11 00:00:00 UTC 2020
Select autophagy genes maintain quiescence of tissue-resident macrophages and increase susceptibility to Listeria monocytogenes.
Nature microbiology
Wang YT,Zaitsev K,Lu Q,Li S,Schaiff WT,Kim KW,Droit L,Wilen CB,Desai C,Balce DR,Orchard RC,Orvedahl A,Park S,Kreamalmeyer D,Handley SA,Pfeifer JD,Baldridge MT,Artyomov MN,Stallings CL,Virgin HW
17-5920 was used in Flow cytometry/Cell sorting to delineate a function of the autophagy proteins Beclin 1 and FIP200-but not of other essential autophagy components ATG5, ATG16L1 or ATG7-in mediating quiescence of tissue-resident macrophages by limiting the effects of systemic interferon-γ.
Sat Feb 01 00:00:00 UTC 2020
Inducing cancer indolence by targeting mitochondrial Complex I is potentiated by blocking macrophage-mediated adaptive responses.
Nature communications
Kurelac I,Iommarini L,Vatrinet R,Amato LB,De Luise M,Leone G,Girolimetti G,Umesh Ganesh N,Bridgeman VL,Ombrato L,Columbaro M,Ragazzi M,Gibellini L,Sollazzo M,Feichtinger RG,Vidali S,Baldassarre M,Foriel S,Vidone M,Cossarizza A,Grifoni D,Kofler B,Malanchi I,Porcelli AM,Gasparre G
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Feb 22 00:00:00 UTC 2019
Irg1 expression in myeloid cells prevents immunopathology during M. tuberculosis infection.
The Journal of experimental medicine
Nair S,Huynh JP,Lampropoulou V,Loginicheva E,Esaulova E,Gounder AP,Boon ACM,Schwarzkopf EA,Bradstreet TR,Edelson BT,Artyomov MN,Stallings CL,Diamond MS
Mon Apr 02 00:00:00 UTC 2018
Chloroquine modulates antitumor immune response by resetting tumor-associated macrophages toward M1 phenotype.
Nature communications
Chen D,Xie J,Fiskesund R,Dong W,Liang X,Lv J,Jin X,Liu J,Mo S,Zhang T,Cheng F,Zhou Y,Zhang H,Tang K,Ma J,Liu Y,Huang B
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Feb 28 00:00:00 UTC 2018
Oncostatin M is overexpressed in skin squamous-cell carcinoma and promotes tumor progression.
Oncotarget
Simonneau M,Frouin E,Huguier V,Jermidi C,Jégou JF,Godet J,Barra A,Paris I,Levillain P,Cordier-Dirikoc S,Pedretti N,Bernard FX,Lecron JC,Morel F,Favot L
17-5920 was used in Flow cytometry/Cell sorting to examine the contribution of oncostatin M to the progression of cutaneous squamous cell carcinoma.
Fri Nov 23 00:00:00 UTC 2018
Oncostatin M is overexpressed in skin squamous-cell carcinoma and promotes tumor progression.
Oncotarget
Simonneau M,Frouin E,Huguier V,Jermidi C,Jégou JF,Godet J,Barra A,Paris I,Levillain P,Cordier-Dirikoc S,Pedretti N,Bernard FX,Lecron JC,Morel F,Favot L
17-5920 was used in Flow cytometry/Cell sorting to examine the contribution of oncostatin M to the progression of cutaneous squamous cell carcinoma.
Fri Nov 23 00:00:00 UTC 2018
Increased Vascular Permeability in the Bone Marrow Microenvironment Contributes to Disease Progression and Drug Response in Acute Myeloid Leukemia.
Cancer cell
Passaro D,Di Tullio A,Abarrategi A,Rouault-Pierre K,Foster K,Ariza-McNaughton L,Montaner B,Chakravarty P,Bhaw L,Diana G,Lassailly F,Gribben J,Bonnet D
17-5920-82 was used in Flow Cytometry to find several abnormalities in the vascular architecture and function in patient-derived xenografts (PDX), such as vascular leakiness and increased hypoxia.
Mon Sep 11 00:00:00 UTC 2017
Mitochondrial permeabilization engages NF-κB-dependent anti-tumour activity under caspase deficiency.
Nature cell biology
Giampazolias E,Zunino B,Dhayade S,Bock F,Cloix C,Cao K,Roca A,Lopez J,Ichim G,Proïcs E,Rubio-Patiño C,Fort L,Yatim N,Woodham E,Orozco S,Taraborrelli L,Peltzer N,Lecis D,Machesky L,Walczak H,Albert ML,Milling S,Oberst A,Ricci JE,Ryan KM,Blyth K,Tait SWG
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Sep 01 00:00:00 UTC 2017
The pattern recognition receptor, Mincle, is essential for maintaining the M1 macrophage phenotype in acute renal inflammation.
Kidney international
Lv LL,Tang PM,Li CJ,You YK,Li J,Huang XR,Ni J,Feng M,Liu BC,Lan HY
17-5920-82 was used in Flow cytometry/Cell sorting to investigate the role of Mincle (macrophage-inducible C-type lectin, Clec4e) in kidney disease.
Wed Mar 01 00:00:00 UTC 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
TGF-β receptor 1 inhibition prevents stenosis of tissue-engineered vascular grafts by reducing host mononuclear phagocyte activation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Lee YU,de Dios Ruiz-Rosado J,Mahler N,Best CA,Tara S,Yi T,Shoji T,Sugiura T,Lee AY,Robledo-Avila F,Hibino N,Pober JS,Shinoka T,Partida-Sanchez S,Breuer CK
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Jul 01 00:00:00 UTC 2016
TNF-Mediated Restriction of Arginase 1 Expression in Myeloid Cells Triggers Type 2 NO Synthase Activity at the Site of Infection.
Cell reports
Schleicher U,Paduch K,Debus A,Obermeyer S,König T,Kling JC,Ribechini E,Dudziak D,Mougiakakos D,Murray PJ,Ostuni R,Körner H,Bogdan C
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue May 03 00:00:00 UTC 2016
Dendritic cell-derived nitric oxide inhibits the differentiation of effector dendritic cells.
Oncotarget
Si C,Zhang R,Wu T,Lu G,Hu Y,Zhang H,Xu F,Wei P,Chen K,Tang H,Yeretssian G,Xiong H
17-5920 was used in Flow cytometry/Cell sorting to demonstrate that DC-derived nitric oxide restrains effector DC development to treat autoimmune and inflammatory diseases.
Tue Nov 15 00:00:00 UTC 2016
Commensal Gram-positive bacteria initiates colitis by inducing monocyte/macrophage mobilization.
Mucosal immunology
Nakanishi Y,Sato T,Ohteki T
Thu Jan 01 00:00:00 UTC 2015
Expression of Cationic Amino Acid Transporter 2 Is Required for Myeloid-Derived Suppressor Cell-Mediated Control of T Cell Immunity.
Journal of immunology (Baltimore, Md. : 1950)
Cimen Bozkus C,Elzey BD,Crist SA,Ellies LG,Ratliff TL
17-5920 was used in Flow cytometry/Cell sorting to investigate whether restricting myeloid-derived suppressor cell uptake of l-Arginine is a critical control point to modulate their suppressor activity.
Tue Dec 01 00:00:00 UTC 2015
Maternal immune activation evoked by polyinosinic:polycytidylic acid does not evoke microglial cell activation in the embryo.
Frontiers in cellular neuroscience
Smolders S,Smolders SM,Swinnen N,Gärtner A,Rigo JM,Legendre P,Brône B
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Aug 24 00:00:00 UTC 2015
Myeloid cell-derived inducible nitric oxide synthase suppresses M1 macrophage polarization.
Nature communications
Lu G,Zhang R,Geng S,Peng L,Jayaraman P,Chen C,Xu F,Yang J,Li Q,Zheng H,Shen K,Wang J,Liu X,Wang W,Zheng Z,Qi CF,Si C,He JC,Liu K,Lira SA,Sikora AG,Li L,Xiong H
17-5920 was used in Flow cytometry/Cell sorting to show that iNOS-deficient mice display enhanced classically activated M1 macrophage polarization without major effects on alternatively activated M2 macrophages.
Fri Mar 27 00:00:00 UTC 2015