Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQLAQKEQE LARAKEALQA MKADRKRLKG EKTDLVSQMQ QLYATLESRE EQLRDFIRNY EQHRKESEDA VKALAKEKDL LEREKWELRR QAKEATDHAT ALRSQLDLKD NRMKELEAEL AMAKQSLATL TKDVPKRHSL AMPGETVLNG NQEWVVQADL PLTAAIRQSQ QTLYHSHPPH PADRQAVRVS PCHSRQPSVI SDASAAEGDR SSTPSDINSP RHRTHSLCNG DSPGPVQKNL HNPIVQSLED LEDQKRKKKK EKMGFGSISR VFARGKQRKS LDPGLFDGTA PDYYIEEDAD W (116-415 aa encoded by BC035501)
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kazrin
Gene Aliases: 2310007B04Rik; 9030409G11Rik; AI256607; AI666764; AW061099; C1orf196; HRIHFB2003; KAZ; KAZN; Kazrin; KIAA1026
UniProt ID: (Human) Q674X7, (Mouse) Q69ZS8, (Rat) Q5FWS6
Entrez Gene ID: (Human) 23254, (Mouse) 71529, (Rat) 313672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support