Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member A; glutamate receptor; glutamate receptor, metabotropic 1; GRM1A; Metabotropic glutamate receptor 1; mGluR1; mGluR1a; protein phosphatase 1, regulatory subunit 85
Gene Aliases: 4930455H15Rik; ENSMUSG00000075319; Gm10828; GPRC1A; GRM1; MGLU1; MGLUR1; nmf373; PPP1R85; rcw; SCAR13; wobl
UniProt ID: (Human) Q13255, (Mouse) P97772, (Rat) P23385
Entrez Gene ID: (Human) 2911, (Mouse) 14816, (Rat) 24414
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support