Search Thermo Fisher Scientific
This antibody is specific for the ~125k monomer or the ~250k dimers of mGluR5 or mGluR1. Immunolabeling can be blocked by preadsorption of antibody with the antigenic peptide. A suggested positive control for this product is HEK-293 cell lysate.
may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.