Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs and Western blots, no cross-reactivity with recombinant human mGluR1, R2, R3, R4, R7, or R8 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
GRK1 is a member of the guanine nucleotide protein-coupled (G protein) receptor kinase subfamily of Serine/Threonine kinase family. GRK1 phosphorylates rhodopsin and initiates its deactivation. G protein mediated phosphorylation promotes the binding of arrestin, which removes the receptor from the G-protein and terminates the receptor signal chain. GRK1 is farnesylated to promote membrane association. Studies have shown that GRK1 has been shown to be inhibited by recoverin, a Calcium binding protein. Diseases associated with GRK1 protein dysfunction include Oguchi Disease-2 and Hereditary Night Blindness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A830007B11Rik; D16Ium24; D16Ium24e; Glur-5; Glur5; Glurbeta1; glutamate receptor; glutamate receptor, metabotropic 5; GPRC1A; GRM 1; GRM 5; GRM1; GRM1 alpha; GRM1A; metabotropic 1; Metabotropic glutamate receptor 5; MGC25101; mGlu1; mGluR; mGluR1 alpha; mGluR1A; mGluR5; mGluR5a; protein phosphatase 1, regulatory subunit 86
Gene Aliases: GPRC1E; GRM5; mGlu5; MGLUR5; PPP1R86
UniProt ID: (Human) P41594
Entrez Gene ID: (Human) 2915
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support