Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31752 has been tested in indirect ELISA.
Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutamate receptor; glutamate receptor, metabotropic 6; Metabotropic glutamate receptor 6; mGluR6
Gene Aliases: CSNB1B; GPRC1F; GRM6; mGlu6; MGLUR6
UniProt ID: (Human) O15303
Entrez Gene ID: (Human) 2916
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support