Search Thermo Fisher Scientific
Red Fluorescent Protein (RFP) is isolated from sea anemone (Discosoma sp, Uniprot ID: Q9U6Y8), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications. DsRed2 (Monomer): Excitation: 584 nm; Emission 607 nm.
Protein Aliases: DsRed; mRFP; mRFP1; Red fluorescent protein; RFP monomer