Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFEDKPHAE GAAVVAAAGE ALQALCQELN LDEGSAAEAL DDFTAIRGNY SLEGEVTHWL ACSLYVACRK SIIPTVGKGI MEGNCVSLTR ILRSAKLSLI QFFSKMKKWM DMSNLPQEFR ERIERLERNF EVSTVIFKKY EPIFLDIFQN PYEEPPKLPR SRKQRRIPCS VKDLFNFCWT LFVYTKGNFR MIGDDLVNSY HLLLCCLDLI FANAIMCPNR QDLLNPSFKG LPSDFHTADF TASEEPPCII AVLCELHDGL LVEAKGIKEH YFKPYISKLF DRKILKGECL LDLSSFTDNS KAVNKEYEEY VLTVGDFDER IFLGADAEEE IGTPRKFTRD TPLGKLTAQA NV (1-351 aa encoded by BC032247)
RB-like 1 is similar in sequence and possibly function to the product of the retinoblastoma 1 (RB1) gene. The RB1 gene product is a tumor suppressor protein that appears to be involved in cell cycle regulation, as it is phosphorylated in the S to M phase transition and is dephosphorylated in the G1 phase of the cell cycle. Both the RB1 protein and RB-like 1 can form a complex with adenovirus E1A protein and SV40 large T-antigen, with the SV40 large T-antigen binding only to the unphosphorylated form of each protein. In addition, both proteins can inhibit the transcription of cell cycle genes containing E2F binding sites in their promoters. Due to the sequence and biochemical similarities with the RB1 protein, it is thought that RB-like 1 may also be a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 107 kDa retinoblastoma-associated protein; MGC40006; p107; pRb1; retinoblastoma-like 1; Retinoblastoma-like protein 1
Gene Aliases: AW547426; CP107; p107; PRB1; RBL1
UniProt ID: (Human) P28749, (Mouse) Q64701, (Rat) D3ZS28
Entrez Gene ID: (Human) 5933, (Mouse) 19650, (Rat) 680111
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support