Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 12
Keep as concentrated solution.
Predicted reactivity: Cat (84%), Sheep (82%), Rhesus Monkey (98%), Bovine (83%).
Positive Control: HeLa, HCT116 (30 µM Cisplatin treatment for 24 hr), HCT116 (30 µM Cisplatin treatment for 48 hr).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protein p21 is a tumor suppressor protein which is an inhibitor of Cyclin-dependent kinases (CDK) and is transcriptionally activated by p53. P21 is important in the response of cells to genotoxic stress and a major transcriptional target of p53 protein. The occurrence of p21 in the nucleus executes binding and inhibition activity of cyclin dependent kinases Cdk1 and Cdk2, and blocks the transition from G1 phase into S phase or from G2 phase into mitosis after DNA damage, which enables the repair of damaged DNA. In the cytoplasm, p21 protein has an anti-apoptotic effect. P21 is able to bind to and inhibit caspase 3, as well as the apoptotic kinases ASK1 and JNK. P21 exhibits a dual function in carcinogenesis, and acts as a tumor suppressor, prevents apoptosis, and acts as an oncogene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases:
Activating Fragment 1; CDK-interacting protein 1; CDK-interaction protein 1; CDNK1A; Cip1); Cyclin-dependent kinase inhibitor 1; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA-6; melanoma differentiation associated protein 6; Melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21
Gene Aliases: CAP20; CDKI; CDKN1; CDKN1A; CIP1; MDA-6; MDA6; P21; p21CIP1; p21WAF; PIC1; SDI1; WAF1
UniProt ID: (Human) P38936, (Mouse) P39689
Entrez Gene ID: (Human) 1026, (Mouse) 12575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support