Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: HeLa cell lysate.
This antibody detects three bands within the 34 kDa region corresponding to p34 and its cleavage products by Western blot.
CDK1 or Cell Division Control protein 1 is essential for the completion of START, the controlling event in the cell cycle that is required to initiate mitosis. CDK1 is a catalytic subunit of a protein kinase complex, called the M-Phase Promoting Factor that induces entry into mitosis and is universal among eukaryotes. Phosphorylation of Bcl-2 in G2/M phase-arrested cells following photodynamic therapy with hypericin involves a CDK1-mediated signal and delays the onset of apoptosis. Therapeutic potential of CDK inhibitor NU2058 in androgen-independent prostate cancer has also been demonstrated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABL1; CDK1; CDK1-A; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; Cell division control protein 2 homolog; Cell division control protein 2 homolog 1; Cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 1-A; Cyclin-dependent kinase 1; Cyclin-dependent kinase 1-A; DKFZp686L20222; maturation-promoting factor; MGC111195; MGC53829; OTTHUMP00000019660; P34 CDK1; P34 MGC111195; p34 protein kinase; p34 protein kinase 1
Gene Aliases:
cdc-2; CDC2; cdc2-a; CDC28A; Cdc2a; cdc2x1.1; CDK1; cdk1-a; cdk1-b; CDKN1; MPF; p34
UniProt ID: (Bovine) P48734, (Human) P06493, (Rat) P39951, (Mouse) P11440, (Xenopus) P35567
Entrez Gene ID: (Bovine) 281061, (Human) 983, (Rat) 54237, (Mouse) 12534, (Xenopus) 380246
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support