Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALQGISVM ELSGLAPGPF CAMVLADFGA RVVRVDRPGS RYDVSRLGRG KRSLVLDLKQ PRGAAVLRRL CKRSDVLLEP FRRGVMEKLQ LGPEILQREN PRLIYARLSG FGQSGSFCRL AGHDINYLAL SGGRNS (1-135 aa encoded by B C009471)
P504S/AMACR/a-Methylacyl-CoA Racemase is an essential enzyme in the b-oxidation of branched-chain fatty acids. High expression of P504S protein is found in prostatic adenocarcinoma but not in benign prostate tissue by immunohistochemical staining in paraffin-embedded tissue. The expression of P504S is also detected in two premalignant lesions of the prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; amcr; OTTHUMP00000219978; OTTHUMP00000219979
Gene Aliases: AMACR; AMACRD; CBAS4; Macr1; RACE; RM
UniProt ID: (Human) Q9UHK6, (Mouse) O09174
Entrez Gene ID: (Human) 23600, (Mouse) 17117
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support