Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive test controls include: HepG2, A-549, BxPC-3, RAW264.7, C6, Mouse lung, Mouse liver, Rat lung.
Immunogen sequence: PPPCFTAGGL TTIKVETSPM ENLLIEHPSM SVYAVHNSCP GLSEATRGTD ELHSPSSPRV EAQNEMGQHI HCYVAALAAH TTFLEQPKSF RPSQWIKEHS E
A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p53-dependent damage-inducible nuclear protein 1; p53-inducible p53DINP1; p53DINP1; stress induced protein; Stress-induced protein; TEAP; thymus expressed acidic protein; Thymus-expressed acidic protein; transformation related protein 53 inducible nuclear protein 1; Tumor protein p53-inducible nuclear protein 1
Gene Aliases: 2700057G22Rik; P53DINP1; SIP; SIP18; SIP27; Stinp; Teap; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; Trp53inp1
UniProt ID: (Human) Q96A56, (Rat) Q80YE2, (Mouse) Q9QXE4
Entrez Gene ID: (Human) 94241, (Rat) 297822, (Mouse) 60599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support