Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Suggested positive control: antigen standard for TP73 (transient overexpression lysate), p73 transfected cells..
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; RP23-254N4.10; Tumor protein p73
Gene Aliases: deltaNp73; P73; TAp73; TP73; Trp73
UniProt ID: (Human) O15350, (Mouse) Q9JJP2
Entrez Gene ID: (Human) 7161, (Mouse) 22062
Molecular Function:
P53-like transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support