Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Cation channel with high affinity for sodium, which is gated by extracellular protons and inhibited by the diuretic amiloride. ASIC1 is permeable for Ca(2+), Li(+) and K(+) and is expressed in dorsal root ganglia, sciatic nerves, olfactory bulb, neo and allocortical regions, dentate granule cells, pyramidal cells of CA1-CA3 subfields of the hippocampal formation, habenula, basolateral amygdaloid nuclei, and in the Purkinje and granule cells of the cerebellum. It is diffusely detected over most other regions of the basal ganglia, including thalamic nuclei, substantia nigra, striatum and globus pallidus, hypothalamus, midbrain, pons, medulla and choroid plexus. Isoform 3 is expressed only in dorsal root ganglion while isoform 1 is expressed in DRG, spinal chord, trigeminal ganglia and the trigeminal mesencephalic nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid sensing (proton gated) ion channel 1; acid sensing (proton gated) ion channel 2; acid sensing (proton gated) ion channel 3; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel 2; acid sensing ion channel 3; acid sensing ion channel family member 4; acid-sensing (proton-gated) ion channel 1; acid-sensing (proton-gated) ion channel 2; acid-sensing (proton-gated) ion channel 3; acid-sensing (proton-gated) ion channel family member 4; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; Acid-sensing ion channel 2; Acid-sensing ion channel 3; Acid-sensing ion channel 4; Amiloride-sensitive brain sodium channel; Amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel 2, neuronal; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; Amiloride-sensitive cation channel neuronal 1; ASIC1; ASIC2; ASIC3; ASIC4; BNaC2; BNC1; Brain sodium channel 1; Brain sodium channel 2; brain sodium channel 4; Cation channel, amiloride-sensitive, neuronal, 2; degenerin; degenerin 2; Dorsal root ASIC; DRASIC; hTNaC1; Mammalian degenerin homolog; MDEG; modulatory subunit of ASIC2a; neuronal amiloride-sensitive cation channel 1; Neuronal amiloride-sensitive cation channel 3; proton gated cation channel ASIC1; proton gated cation channel DRASIC; proton-gated cation channel subunit; putative acid-sensing ion channel; spinal chord ASIC; Spinal cord ASIC; Testis sodium channel 1
Gene Aliases: ACCN; ACCN1; ACCN2; ACCN3; ACCN4; ACIC2; AI843610; ASIC; ASIC1; ASIC1a; ASIC2; ASIC2a; ASIC3; ASIC4; AW742291; B530003N02Rik; BNAC1; BNaC1a; BNAC2; BNAC4; BNC1; BNC1k; DRASIC; hBNaC1; MDEG; MDEG1; MDEG2; SLNAC1; Spasic; TNAC1
UniProt ID: (Human) P78348, (Human) Q16515, (Human) Q9UHC3, (Human) Q6PIN9, (Mouse) Q6NXK8, (Rat) P55926, (Rat) Q62962, (Mouse) Q925H0, (Mouse) Q6X1Y6, (Rat) O35240, (Rat) Q9JHS6, (Mouse) Q7TNS7
Entrez Gene ID: (Human) 41, (Human) 40, (Human) 9311, (Human) 55515, (Mouse) 11419, (Rat) 79123, (Rat) 25364, (Mouse) 11418, (Mouse) 171209, (Rat) 286920, (Rat) 63882, (Mouse) 241118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support