Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratins 1, 4, 5, 6, and 8 are members of the type II neutral to basic subfamily. Antibody to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. Anti Pan Cytokeratin (mixture) is a broadly reactive reagent, which recognizes epitopes present in most human epithelial tissues. It facilitates typing of normal, metaplastic and neoplastic cells. Synergy between the various components results in staining amplification. This enables identification of cells, which would otherwise be stained only marginally. The mixture may aid in the discrimination of carcinomas and nonepithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micrometastases in lymph nodes, bone marrow and other tissues and for determining the origin of poorly differentiated tumors. There are two types of cytokeratins the acidic type I cytokeratins and the basic or neutral type II cytokeratins. Cytokeratins are usually found in pairs comprising a type I cytokeratin and a type II cytokeratin. Usually the type II cytokeratins are 8kD larger than their type I counterparts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40-kDa keratin intermediate filament; Cell proliferation-inducing gene 46 protein; CK-18; CK-19; CK-42; cytokeratin 18; cytokeratin 19; Cytokeratin endo B; Cytokeratin-18; Cytokeratin-19; Cytokeratin-42; Endo B; K18; K19; K42; keratin 17n; keratin 18, type I; keratin 19, type I; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; Keratin D; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; Keratin, type I cytoskeletal 42; keratin, type I, 40-kd; Keratin-17n; Keratin-18; Keratin-19; Keratin-42; krt18 {ECO:0000250|UniProtKB:P05783}; krt42; krt42 {ECO:0000250|UniProtKB:Q6SEK1}; predicted ortholog of type I keratin KA22; Type I keratin Ka19; Type I keratin Ka22
Gene Aliases: 2410039E07Rik; AI663979; CK-18; CK18; CK19; CYK18; ecat6; EndoC; K17n; K18; K19; K1CS; Ka19; Ka22; Kerd; Krt-1.19; Krt1-18; Krt1-19; KRT18; KRT19; Krt42; PIG46
UniProt ID: (Human) P05783, (Human) P08727, (Rat) Q5BJY9, (Mouse) P05784, (Mouse) P19001, (Rat) Q63279, (Rat) Q6IFU7, (Mouse) Q6IFX2
Entrez Gene ID: (Human) 3875, (Human) 3880, (Rat) 294853, (Mouse) 16668, (Mouse) 16669, (Rat) 360626, (Rat) 450231, (Mouse) 68239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support