Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Immunogen sequence: ITAHYTLNR QAMGELSELS FHYLIMAKGV IVRSGTHTLP VESGDMKGSF ALSFPVESDV APIARMFIFA ILPDGEVVGD SEKFEIENCL ANKVDLSFSP AQSPPASHAH LQVAAAPQSL CALRAVDQSV LLMKPEAELS VSSVYNLLTV KDLTNFPDNV DQQEEEQGHC PRPFFIHNGA IYVPLSSNEA DIYSFLKGMG LKVFTNSKIR KPKSCSVIPS VSAGAVGQGY YGAGLGVVER PYVPQLGTYN VIPLNNEQSS GPVPETVRSY FPETWIWELV AVNSSGVAEV GVTVPDTITE WKAGAFCLSE DAGLGISSTA SLRAFQPFFV ELTMPYSVIR GEVFTLKATV L (341-690 aa encoded by BC111756)
The protein encoded by this gene is highly expressed in late-pregnancy serum and is similar in structure to alpha-2-macroglobulin. The encoded protein, which acts as a homotetramer, inhibits the activity of all four classes of proteinases. This protein contains cleavage sites for several proteinases. Upon binding of a proteinase, the conformation of this protein changes to trap the proteinase, limiting its activity. This protein appears to be elevated in the sera of presymptomatic Alzheimer's disease patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6; Pregnancy zone protein; pregnancy-zone protein
Gene Aliases: CPAMD6
Entrez Gene ID: (Human) 5858
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support