Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Essential bifunctional enzyme that catalyzes both the N-deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Plays a role in diffusion of morphogen wingless (wg) via its role in heparan sulfate proteoglycans (HSPGs) biosynthesis, HSPGs being required for movement of wg morphogens. Required for wg signaling during both embryonic and imaginal disk development. Also required for FGF receptor signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase; CG8339-PA; CG8339-PB; diet-wingless; Glucosaminyl N-deacetylase/N-sulfotransferase; sfl-PA; sfl-PB; Sulfateless
Gene Aliases: CG8339; CT24567; Dmel\CG8339; Dmel_CG8339; dw; l(3)03844; lincRNA.S4253; NDST; Sfl
UniProt ID: (Fruit fly) Q9V3L1
Entrez Gene ID: (Fruit fly) 38736
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support