Screen, Confirm and Quantify – Q Exactive Orbitrap MS
High-resolution accurate-mass LC-MS instruments for emerging needs
Thermo Scientific hybrid quadrupole-Orbitrap mass spectrometers enable both targeted and non-targeted screening and characterization. They also work with a broad range of sample type applications so that your laboratory can go from compound discovery to quantitation without relying on different instruments and technologies. With the combination of scan speed, high resolution, mass accuracy, spectral quality, and sensitivity you can achieve quantifiable certainty.
Contact us   Grant writing support   FDA NDMA method
On this page
Genotoxic impurity analysis for NDMA in drug products using Orbitrap LC-MS
The Food and Drug Administration (FDA) released a liquid chromatography-high resolution mass spectrometry (LC-HRMS) method for the quantitation of N-nitrosodimethylamine (NDMA) in Ranitidine drug substance and drug products using the Q Exactive hybrid quadrupole-Orbitrap mass spectrometer.
More versatility
Q Exactive Plus Hybrid
Quadrupole-Orbitrap
Mass Spectrometer
Move from application to application with ease.
Learn more
High-resolution native MS
Q Exactive UHMR Hybrid
Quadrupole-Orbitrap
Mass Spectrometer
Improve disease understanding and therapeutic effectiveness.
Learn more
Inside the Q Exactive Hybrid Quadrupole-Orbitrap
  1. RF lens: The radio frequency (RF) lens stacked-ring RF ion guide captures and focuses ions into a tight beam after they exit the ionization source, increasing sensitivity. Large variable spacing between electrodes enables better pumping efficiency and improved ruggedness. Bent flatapole ion guide reduces noise by preventing neutrals and high-velocity clusters from entering the quadrupole.
  2. AABG: Reducing noise and extending maintenance intervals, the Advanced Active Beam Guide (AABG) enables intelligent ion beam management for high flux ion sources.
  3. Quadrupole Mass Filter with AQT: The mass filter improves precursor selection and ion transmission for more accurate quantitation of low abundance analytes in complex matrices. It also offers variable precursor ion isolation width selection from 0.4 Da to full mass range.
  4. Orbitrap mass analyzer: The Orbitrap mass analyzer offers unprecedented high resolution accurate mass (HRAM) analysis of ions as a function of their rotational frequencies; these data are converted to mass spectra using fast Fourier transforms (FFTs).
  5. HCD cell: Ions pass from the nitrogen-filled C-trap into this multipole collision cell and undergo fragmentation. Afterwards, HCD voltage is increased and the ions are transferred back to the C-trap, to be injected into the Orbitrap for detection.
Which Q Exactive Orbitrap Mass Spectrometer is best for my lab?
Our hybrid quadrupole-Orbitrap mass spectrometry systems vary in terms of mass resolution, scan speed and analysis modes. Ideal for mass spectral analysis for targeted and high-throughput analysis of low-level components in complex matrices applications in biopharma, forensic toxicology, clinical and translational research, environmental and food testing. To find the system best suited to your laboratory’s specific needs, compare Q Exactive Orbitrap mass spectrometers using data in the chart below.
 Q Exactive PlusQ Exactive UHMR
 Q Exactive
Plus
Q Exactive
UHMR
Ideal ApplicationsForensic Toxicology, Clinical Research, Food & Environmental Safety Testing, Basic Academic Research, Sport Anti-Doping, Biopharma development, Metabolomics, LipidomicsProteomics, Structural Biology
AnalysesMS and MS/MS, Qual/Quan, Screening & ConfirmationMS and MS/MS, Screening & Confirmation
Standard Features
Resolving Power140,000
(up to 280,000 with BioPharma option)
200,000 at m/z 400
Mass Accuracy
(Internal Calibration)
<1 ppm with internal calibration
Mass Rangem/z 50 - 6,000
(up to 8000 with option)
m/z 350 - 80,000
Scan SpeedUp to 12 HzUp to 12 Hz (at resolution setting of 12,500 @ m/z 400
Ion SourceIonMax with H-ESI II probe(1 full cycle <1 sec)
Polarity Switching1 full cycle <1 sec at 35,000 resolution settingIonMax with H-ESI II probe
Multiplexing10N/A
Scan FunctionsSIM & PRMFS, AIF, MS/MS, pseudo-MS3
DissociationIn-source CID, HCDIn-source trapping, HCD
Optional Upgrades
Intact Protein ModeYes
High Mass Rangen(HMRn, up to 8000 m/z)YesN/A
Enhanced Resolution ModeYes
Performance and Value
Keep up with the speed of science the lab-forward trade-in program
Keep up with the speed of science
The Lab-Forward Trade-In Program
The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-In Program. For a limited time, trade in your chromatography and mass spectrometry instruments and save!
Learn more
High Resolution Mass Spectrometry with Orbitrap Mass Analyzer for Screening, Confirmative and Quantitative Analyses
Learn how Orbitrap mass analyzer instruments enable consistent analytical certainty even in the most complex matrices based on their high and ultra-high mass resolution capabilities, high sensitivity and selectivity in full scan (and MS/MS) acquisition, mass accuracy and precision (robustness) and ease of use.
Download white paper
Laboratories performing LC-MS applications strive for certainty when identifying detected compounds. For many small molecule applications, conventional approaches aren’t enough. Laboratories require more, such as rich high quality spectral information and fit for purpose data analysis software to make satisfactory identifications and characterization.
Compound Discoverer software helps you extract high-confidence understanding from comprehensive Q Exactive high-resolution accurate-mass data.
Q Exactive Orbitrap MS ordering guide
Nitrosamine impurity in pharmaceuticals drugs - Stefano Maggi Interview 2020
With the FDA enforcing the recall of all ranitidine products, the analysis of nitrosamines particularly NDMA has never been more critical.
Video Player is loading.
Current Time 0:00
Duration 2:51
Loaded: 0%
Stream Type LIVE
Remaining Time 2:51
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextNitrosamine impurity in pharmaceuticals drugs - Stefano Maggi Interview 2020
        With the FDA enforcing the recall of all ranitidine products, the analysis of nitrosamines particularly NDMA has never been more critical.
      2. Now Playing
        Up NextNitrosamine impurity in pharmaceuticals drugs found in products of various pharmaceutical vendors​
        With the FDA enforcing the recall of all ranitidine products, the analysis of nitrosamines particularly NDMA has never been more critical.
      3. Now Playing
        Up NextQ Exactive UHMR Mass Spectrometer Product Video
        In this video - Prof. Albert Heck, University of Utrecht, and Alexander Makarov, Thermo Fisher Scientific, discuss the developing field of native MS.
      4. Now Playing
        Up NextOrbitrap MS - Built for BioPharma
        Prof. Alexander Makarov takes us through the evolution of Obitrap technology for the analysis of biotherapeutics
      5. Now Playing
        Up NextThermo Fisher Scientific RadEye™ SPRD Personal Radiation Detector - Overview with Trent North
        Trent North gives an SPRD overview from IACP 2018
    Successful Grant-Writing Starts Here
    7 steps to your grant-writing success
    A well-thought-out and detailed grant proposal is essential. Our technical experts are with you every step of the way in the grant-writing process to help you reach new heights.
    Download infographic 
    LC-MS webinar series
    Expand your application knowledge
    Our LC-MS on-demand webinar series expands your application knowledge for pharma, biopharma, environmental, omics, food, forensics and clinical research and more. Learn how the latest high-performance mass spectrometry solutions can enable your laboratory to solve your most pressing analytical challenges, regardless of sample type.