Oligonucleotides are the starting point for many of today’s biology research, drug discovery, and diagnostics applications. These highly technical applications demand high-quality oligonucleotides for success. Our oligos are made to your specifications with rigorous quality control and quick turnaround for use in a variety of applications, including PCR, cloning, sequencing, and gene detection.
Explore the easy-to use ordering portals for your tube and plate oligos, our complex and large-scale oligo services, and additional resources below.
Available custom oligo formats
 Standard oligos in tubesStandard oligos in platesComplex and large-scale oligos
Oligo icon
Order now
24-well plate icon
Order now
Special request icon
Special requests
Scales25 nmol, 50 nmol, 200 nmol, 1 µmol, 10 µmolMilligram to >15 gram final yield
LengthsUp to 100 bases
PurificationDesalted, cartridge, HPLC, PAGEDesalted, HPLC, RP-HPLC, IEX-HPLC, Dual-HPLC
Modifications5’, internal, 3’
Formats
  • Tube 
  • Dried oligo shipped ambient
  • Oligo resuspended in water shipped ambient or frozen
  • 96- or 384-well plate
  • Dried oligo shipped ambient 
  • Oligo resuspended in TE, Tris, or water shipped ambient or frozen
Custom formulations and packaging solutions
OrderingEnter individually or bulk upload up to 200 sequences at onceUpload each plate separately using our template
  • Submit a request to our dedicated technical support team
  • Commercial supply available
ShippingStandard or next-day delivery, with free overnight shipping*Standard delivery, with free overnight shipping* 
Additional resourcesModification and configuration options
Online ordering guide
Download 96-well template
Download 384-well template
Complex and large-scale capabilities
All oligos are manufactured using controlled procedures certified against ISO 9001:2015 and ISO 13485:2016 standards.
Custom DNA oligos brochure
For a comprehensive guide on the small- and large-scale custom solutions and end-to-end services designed with your unique application in mind, access the Custom DNA oligos brochure.
 Download brochure
Our custom oligonucleotide synthesis process
Our custom DNA oligos are synthesized on a highly automated, computer-controlled system using standard cyano-ethyl phosphoramidite chemistry. Post-synthesis QC employs electrospray ionization mass spectrometry (ESI MS) for all oligos. As a standard practice, 100% of synthesized oligos are measured for quality control whether provided in tubes or in plates.
Video Player is loading.
Current Time 0:00
Duration 1:41
Loaded: 9.74%
Stream Type LIVE
Remaining Time 1:41
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Choose DNA oligos for your application
    While our value oligo format represents our most economical and fastest way to order (25 or 50 nmol in tube formats with a range of purification options to suit your applications), if your research demands a scale larger than 50 nmol, a different format, or requires 5′, 3′, or internal modifications, then our standard DNA oligo formats have you covered (Table 1).
    Table 1. Which kind of oligos are right for me?
    Value oligosStandard oligos
    Price is calculated per oligoPrice is calculated per base
    • Length: 5–40 bases
    • Scale: 25–50 nmol
    • Length: 5–100 bases
    • Scale: from 25 nmol to 10 µmol
    Not available with modifications5′, internal, and 3′ modifications are available
    Order nowOrder now
    Choosing the synthesis scale and purification option that is well suited for your application depends on the nature of your downstream applications. We can help you choose the right oligos and purification methods for your application (Table 2).
    Table 2. Recommended oligo processing options by application
    ApplicationDesaltedCartridgeHPLCPAGE
    • 25 nmole–10 µmole
    • 5–100 bases
    • Removes salts but not truncated sequences
    • 50 nmole–1 µmole
    • 7–55 bases
    • Removes truncated sequences
    • Provides 80% full-length sequence
    • 50 nmole–10 µmole
    • 10–55 bases
    • Removes truncated sequences
    • Provides >85% full-length sequence
    • 50 nmole–10 µmole
    • 7–100 bases
    • Removes truncated sequences
    • Provides > 90% full-length sequence
    Standard PCR    
    Specialty PCR    
    Cloning    
    cDNA library construction    
    Sequencing    
    Fluorescent sequencing    
    Next generation sequencing    
    Mutagenesis    
    Antisense    
    Gel shift assays    
    Microarrays    
    Looking for pre-defined oligo sequences?
    In addition to our custom synthesis options for oligos, primers, probes, and genes, we offer random primers, oligo (dT) primers, 5’ labeled primers, gene specific primers for sequencing, forward and reverse sequencing primers and kits.
    Order now
    Video Player is loading.
    Current Time 0:00
    Duration 4:43
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 4:43
    1x
      • Chapters
      • descriptions off, selected
      • en (Main), selected
      Watch now: Learn how easy it is to order custom oligos online.
      Find everyday lab essentials to power your PCR every step of the way. Shop now
      * This promotion is open to customers in the US (excluding Puerto Rico) and Canada. Discount will apply to qualifying orders received by Life Technologies no later than December 31, 2024. Discount applies to list price in effect at the time order is received by Life Technologies. Cannot be combined with other discounts or promotions. Offer void where prohibited, licensed or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
      Stylesheet for Classic Wide Template adjustments
      For Research Use Only. Not for use in diagnostic procedures.