Proteins interact with DNA through electrostatic interactions (salt bridges), dipolar interactions (hydrogen bonding, H-bonds), entropic effects (hydrophobic interactions) and dispersion forces (base stacking). These forces contribute in varying degrees to proteins binding in a sequence-specific or non–sequence-specific manner. Understanding how proteins interact with DNA, determining what proteins are present in these protein–DNA complexes, and identifying the nucleic acid sequence (and possible structure) required to assemble these complexes are vital to understanding the role these complexes play in regulating cellular processes. A number of laboratory techniques have been developed to study the complex interactions of proteins with DNA, each with a unique history, varying utility, and distinct strengths and weaknesses.
Chromatin immunoprecipitation (ChIP) assays
The chromatin immunoprecipitation (ChIP) method can be used to monitor transcriptional regulation through histone modification (epigenetics) or transcription factor–DNA binding interactions. The ChIP assay method allows analysis of DNA–protein interactions in living cells by treating the cells with formaldehyde or other crosslinking reagents in order to stabilize the interactions for downstream purification and detection. Performing ChIP assays requires knowledge of the target protein and DNA sequence that will be analyzed, as researchers must provide an antibody against the protein of interest and PCR primers for the DNA sequence of interest. The antibody is used to selectively precipitate the protein–DNA complex from the other genomic DNA fragments and protein–DNA complexes. The PCR primers allow specific amplification and detection of the target DNA sequence. Quantitative PCR (qPCR) technique allows the amount of target DNA sequence to be quantified. The ChIP assay is amenable to array-based formats (ChIP-on-chip) or direct sequencing of the DNA captured by the immunoprecipitated protein (ChIP-seq).
StrengthsLimitations 
  • capture a snapshot of specific protein–DNA
    interactions as they occur in living cells
  • quantitative when coupled with qPCR analysis 
  • ability to profile a promoter for different proteins
  • researcher needs to source ChIP-grade antibodies
  • requires designing specific primers
  • difficult to adapt for high-throughput screening
A step-by-step guide to successful chromatin immunoprecipitation (ChIP) assays
This updated overview of the ChIP procedure includes additional detail about primary antibody selection (i.e., ChIP-validated antibodies). The application note also describes and provides examples of chromatin immunoprecipitation (ChIP) as a technique for studying epigenetics, as it allows researchers to capture a snapshot of specific protein–DNA interactions.
Protein Interactions Handbook
Our 72-page Protein Interaction Technical Handbook provides protocols and technical and product information to help maximize results for protein interaction studies. The handbook provides background, helpful hints and troubleshooting advice for immunoprecipitation and co-immunoprecipitation assays, pull-down assays, far-western blotting and crosslinking. The handbook also features an expanded section on methods to study protein–nucleic acid interactions, including ChIP, EMSA, and RNA EMSA. The handbook is an essential resource for any laboratory studying protein interactions.
Contents include: Introduction to protein interactions, Co-immunoprecipitation assays, Pull-down assays, Far-western blotting, Protein interaction mapping, Yeast two-hybrid reporter assays, Electrophoretic mobility shift assays [EMSA], Chromatin immunoprecipitation assays (ChIP), Protein–nucleic acid conjugates, and more.
 Protein Interactions Handbook
DNA electrophoretic mobility shift assay (EMSA)
The DNA electrophoretic mobility shift assay (EMSA) is used to study proteins binding to known DNA oligonucleotide probes and can be used to assess the degree of affinity or specificity of the interaction. The technique is based on the observation that protein–DNA complexes migrate more slowly than free DNA molecules when subjected to non-denaturing polyacrylamide or agarose gel electrophoresis. Because the rate of DNA migration is shifted or retarded upon protein binding, the assay is also referred to as a gel shift or gel retardation assay. Adding a protein-specific antibody to the binding components creates an even larger complex (antibody–protein–DNA), which migrates even slower during electrophoresis. This is known as a “supershift”, and it can be used to confirm protein identities. Until conception of the EMSA, protein–DNA interactions were studied primarily by nitrocellulose filter–binding assays using radioactively labeled probes.
StrengthsLimitations
  • detect low abundance DNA binding proteins from lysates
  • test binding site mutations using many probe configurations with the same lysate
  • test binding affinity through DNA probe mutational analysis
  • non-radioactive EMSA possible using biotinylated or fluorescently labeled DNA probes
  • analyze protein–DNA interactions in vitro
  • difficult to quantitate
  • need to perform supershift assay with antibody to be certain of protein identity in a complex
Traditionally, DNA probes have been radiolabeled with ³²P by incorporating an [γ-³²P]dNTP during a 3' fill-in reaction using Klenow fragment or by 5' end labeling using [γ-³²P]ATP and T4 polynucleotide kinase. Following electrophoresis, the gel is exposed to X-ray film to document the results. The Thermo Scientific LightShift Chemiluminescent EMSA Kit is a non-radioactive assay that provides robust and sensitive performance. The kit includes reagents for setting up and customizing DNA-binding reactions, a control set of DNA and protein extract to test the kit system, stabilized streptavidin–HRP conjugate to probe for the biotin-labeled DNA target, and an exceptionally sensitive chemiluminescent substrate module for detection.
Chemiluminescent EMSA of four different DNA–protein complexes. Biotin-labeled target duplexes ranged in size from 21–25 bp. The Oct-1, AP1 and NF-κB transcription factors were derived from HeLa nuclear extract. EBNA-1 extract is provided as a control in the LightShift Chemiluminescent EMSA Kit. Unlabeled specific competitor sequences (where used) were present at a 200-fold molar excess over labeled target. X-ray film exposure times for each system ranged from 2 minutes for EBNA, Oct-1 and AP1, and 5 minutes for NF-κB.
DNA pull-down assays
Pull-down assays are used to selectively extract a protein–DNA complex from a sample. Typically, the pull-down assay uses a DNA probe labeled with a high affinity tag, such as biotin, which allows the probe to be recovered or immobilized. A biotinylated DNA probe can be complexed with a protein from a cell lysate in a reaction similar to that used in the EMSA and then used to purify the complex using agarose or magnetic beads. The proteins are then eluted from the DNA and detected by western blot or identified by mass spectrometry. Alternatively, the protein may be labeled with an affinity tag, or the DNA–protein complex may be isolated using an antibody against the protein of interest (similar to a supershift assay). In this case, the unknown DNA sequence bound by the protein is detected by Southern blotting or through PCR analysis.
StrengthsLimitations
  • enrichment of low abundant targets
  • end-labeled DNA can be generated by several methods
  • isolation of intact complex
  • compatible with immunoblotting and mass spectrometry analysis
  • long DNA probes can show significant nonspecific binding
  • requires very specific antibodies for native proteins
  • requires nuclease-free conditions
  • assay must be performed in vitro
Microplate capture and detection assays
A hybrid of the DNA pull-down assay and enzyme-linked immunosorbent assay (ELISA), microplate capture assays use immobilized DNA probes to capture specific protein–DNA interactions and confirm protein identities and relative amounts with target specific antibodies. Typically, a biotinylated DNA probe is immobilized on the surface of a 96- or 384-well microplate coated with streptavidin. A cellular extract is prepared in binding buffer and added for a sufficient amount of time to allow the putative binding protein to bind to the oligonucleotide. The extract is then removed and each well is washed several times to remove nonspecifically bound proteins. Finally, the protein is detected using a specific antibody labeled for detection. This method can be extremely sensitive when performed with enzyme-labeled antibodies and a chemiluminescent substrate, detecting less than 0.2 pg of the target protein per well. The microplate format is efficient and compatible with high-throughput analysis, allowing statistical mutational and activation assays to be performed. This method may also be utilized for oligonucleotides labeled with other tags, such as primary amines that can be immobilized on microplates coated with an amine-reactive surface chemistry.
StrengthsLimitations
  • the use of ELISA-based technology increases speed and throughput
  • compatible with drug screening
  • possible to optimize sensitive non-radioactive assays
  • requires antibodies with affinity for DNA-bound native proteins (i.e., supershift antibodies)
  • data only provides relative changes in transcription factor–DNA affinity or abundance
  • assay kits are available for only a few targets
Reporter assays
Reporter assays provide a real-time in vivo readout of translational activity for a promoter of interest. Reporter genes are fusions of a target promoter DNA sequence and a reporter gene DNA sequence. The promoter DNA sequence is customized by the researcher and the reporter gene DNA sequence codes for a protein with detectable properties such as firefly luciferase, Renilla luciferase or alkaline phosphatase. These genes produce enzymes only when the promoter of interest is activated. The enzyme, in turn, catalyzes a substrate to produce either light, a color change, or other reaction that can be detected by spectroscopic instrumentation. The signal from the reporter gene is used as an indirect determinant for the translation of endogenous proteins driven from the same promoter.
StrengthsLimitations
  • in vivo monitoring
  • captures real-time data
  • powerful tool for mutational analysis of promoters
  • amenable to high-throughput screening
  • uses exogenous DNA
  • does not address changes due to genomic sequences 
    near the promoter of interest
  • artifacts due to gene dosage can occur
Watch this video to learn more about luciferase reporter assays
Video Player is loading.
Current Time 0:00
Duration 4:46
Loaded: 3.47%
Stream Type LIVE
Remaining Time 4:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video Player is loading.
    Current Time 0:00
    Duration 5:06
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 5:06
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Video Player is loading.
      Current Time 0:00
      Duration 4:46
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 4:46
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Video Player is loading.
        Current Time 0:00
        Duration 5:06
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 5:06
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Recommended reading
          1. Hendrickson W (1985) BioTechniques 3:346–354.
          2. Evertts AG et al. (2010) Modern approaches for investigating epigenetic signaling pathways. J Appl Physiol Jan 28 Epub ahead of print.
          3. Georges AB et al. (2010) Generic binding sites, deneric DNA-binding domains: Where does specific promoter recognition come from? FASEB Journal 24:346–356.
          4. Lunde BM et al. (2007) RNA-binding proteins: modular design for efficient function. Nat Rev Mol Cell Biol 8:479–490.
          For Research Use Only. Not for use in diagnostic procedures.