Thermo Scientific hydrocarbons
Hydrocarbons are organic compounds made up entirely of carbon and hydrogen atoms. Alkanes are the simplest organic compounds, having only single bonds between carbon atoms. They are referred to as saturated hydrocarbons and have the the general formula of CnH2n+2, where n is the number of carbon atoms. Alkenes have at least one carbon-carbon double bond and have the general formula CnH2n. Alkynes have one or more carbon-carbon triple bonds and have the general formula CnH2n-2. Alkenes and alkynes are called unsaturated hydrocarbons. Alkanes have the general formula of CnH2n+2, where n is the number of carbon atoms.
Hydrocarbons are non-polar compounds and are insoluble in water. They have low boiling and melting points. In general, alkanes and cycloalkanes have low reactivity and are used as solvents in organic chemistry. Natural gas, camping gas, lighter gas, and most gasolines are all alkanes. Alkenes and alkynes are more reactive than alkanes. Unsaturated compounds (with double or triple bonds) form addition and cycloaddition compounds, in which two substances combine to form a single substance.
The Thermo Scientific portfolio includes hydrocarbons suitable for a range of laboratory research applications.
Shop now  Contact us
Alkanes are hydrocarbons that contain only carbon-carbon single bonds.
Catalog # Name Size Price (USD) Qty
309165000 Tetracontane, 98% Each -
042460.18 n-Octacosane, 99% Each
81.65
Special offer
Online exclusive
95.40
Save 13.75 (14%)
A13853.22 n-Eicosane, 99% Each
59.65
Online Exclusive
66.10
Save 6.45 (10%)
031209.22 n-Decane, 99+% Each -
043283.LV n-Hexadecane, 95% Each
154.65
Special offer
Online exclusive
182.00
Save 27.35 (15%)
A14672.0E n-Tetradecane, 99% Each
634.65
Special offer
Online exclusive
746.00
Save 111.35 (15%)
A18198.14 n-Heneicosane, 99% Each
272.65
Special offer
Online exclusive
320.00
Save 47.35 (15%)
031807.18 n-Undecane, 99% Each
50.65
Special offer
Online exclusive
58.90
Save 8.25 (14%)
Alkenes are hydrocarbons that contain at least one carbon-carbon double bond, while alkynes contain at least one carbon-carbon triple bond. Both are more reactive than alkanes.
Catalog # Name Size Price (USD) Qty
A12139.22 Phenylacetylene, 98+% Each
108.65
Special offer
Online exclusive
128.00
Save 19.35 (15%)
H53500.06 3-Phenyl-1-propyne, 97%, stab. Each
104.65
Special offer
Online exclusive
123.00
Save 18.35 (15%)
L11608.06 5-Phenyl-1-pentyne, 98+% Each
72.65
Special offer
Online exclusive
85.10
Save 12.45 (15%)
A12139.14 Phenylacetylene, 98+% Each
41.65
Online Exclusive
46.40
Save 4.75 (10%)
A12139.36 Phenylacetylene, 98+% Each
477.65
Online Exclusive
531.00
Save 53.35 (10%)
H53500.14 3-Phenyl-1-propyne, 97%, stab. Each
394.65
Online Exclusive
438.00
Save 43.35 (10%)
149910010 trans-3-Hexene, 98% Each -
149910051 trans-3-Hexene, 98% Each -
014685.30 Manganese(II) oxalate dihydrate, Mn 30% min Each
209.65
Special offer
Online exclusive
247.00
Save 37.35 (15%)
152461000 Phenylacetylene, 98%, pure Each
111.65
Special offer
Online exclusive
131.00
Save 19.35 (15%)
152460250 Phenylacetylene, 98%, pure Each
48.65
Special offer
Online exclusive
57.20
Save 8.55 (15%)
152465000 Phenylacetylene, 98%, pure Each -
B22636.06 1,3,5,7-Cyclooctatetraene, 98%, stab. with 0.1% Hydroquinone Each
154.65
Special offer
Online exclusive
182.00
Save 27.35 (15%)
B22636.14 1,3,5,7-Cyclooctatetraene, 98%, stab. with 0.1% Hydroquinone Each
536.65
Online Exclusive
596.00
Save 59.35 (10%)
L11608.14 5-Phenyl-1-pentyne, 98+% Each
259.65
Special offer
Online exclusive
305.00
Save 45.35 (15%)
Cyclic hydrocarbons are formed when carbon atoms combine to form a ring. Polycyclic hydrocarbons are made up of two or more fused cyclic hydrocarbons.
Catalog # Name Size Price (USD) Qty
A13883.0D Decahydronaphthalene, cis + trans, 98% Each
531.65
Special offer
Online exclusive
625.00
Save 93.35 (15%)
146110050 Adamantane, 99+% Each
18.65
Special offer
Online exclusive
21.40
Save 2.75 (13%)
A13883.0F Decahydronaphthalene, cis + trans, 98% Each
227.65
Special offer
Online exclusive
268.00
Save 40.35 (15%)
B21953.06 1-Phenylnaphthalene, 97% Each
62.65
Online Exclusive
69.40
Save 6.75 (10%)
B21953.14 1-Phenylnaphthalene, 97% Each
194.65
Special offer
Online exclusive
229.00
Save 34.35 (15%)
B21953.22 1-Phenylnaphthalene, 97% Each
702.65
Special offer
Online exclusive
826.00
Save 123.35 (15%)
A13883.AE Decahydronaphthalene, cis + trans, 98% Each
36.65
Online Exclusive
40.90
Save 4.25 (10%)
406171000 Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™ Each -
406170010 Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™ Each
127.65
Special offer
Online exclusive
150.00
Save 22.35 (15%)
A15327.36 1-Methylnaphthalene, 96% Each
84.65
Special offer
Online exclusive
99.80
Save 15.15 (15%)
146110250 Adamantane, 99+% Each
37.65
Special offer
Online exclusive
43.60
Save 5.95 (14%)
A13883.AP Decahydronaphthalene, cis + trans, 98% Each
93.65
Online Exclusive
104.00
Save 10.35 (10%)
A15327.22 1-Methylnaphthalene, 96% Each
31.40
146111000 Adamantane, 99+% Each -
146110010 Adamantane, 99+% Each -
In addition to our catalog range shown above, we can deliver bulk quantities of various hydrocarbon products to suit your custom or bulk requirements.
Learn more