Visualize your imaging data and obtain qualitative and quantitative information such as volume, density and diameter, from a single software environment.
Anatomical image analysis relies on augmenting structural information from micro-CT or MRI systems with functional data from PET, SPECT or optical imaging instrumentation. Analysis of novel drug candidates requires pinpointing the location of dynamic processes acquired with functional modalities in images of the animal’s anatomy from structural modalities over time.
Amira Software enables the fusion of any type of anatomical image data. Co-registered images can be segmented and analyzed using Amira Software’s vast library of image analysis filters and algorithms. Image segmentation and quantification workflows can be applied to entire time series of image data of dynamic processes. With Amira Software’s professional-grade visualization tools, structural information from CT and MRI can be augmented and rendered jointly with segmentation results and functional images to create visualization of physiological processes that reveal the underlying mechanisms in an intuitive animation.
Video Player is loading.
Current Time 0:00
Duration 1:19
Loaded: 3.77%
Stream Type LIVE
Remaining Time 1:19
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Imaging data
    Workflow using Avizo Software
    Processing
    Workflow using Avizo Software
    Visualization
    Workflow using Avizo Software
    Analysis
    Workflow using Avizo Software
    Customization
    Confidence
    Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to biomedical and life science research.
    Customization
    Customization
    Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
    icon-improve_collaboration_64x64
    Support
    Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
    Software_Automation_65x64
    Automation
    Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency.
    Scientific use cases using Amira Software
    Testimonials
    Video Player is loading.
    Current Time 0:00
    Duration 5:10
    Loaded: 1.65%
    Stream Type LIVE
    Remaining Time 5:10
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      VetCore, University of Veterinary Medicine uses Amira Software
      "Research at Vetmeduni Vienna is quite diverse, covering both clinical and preclinical research areas. We are currently four people working in the imaging facility, providing equipment and supports for researchers in both image acquisition (widefield microscopy, CLSM, spinning disk confocal microscopy, soon also STORM super-resolution microscopy, microscopic X-ray computed tomography) and in data visualization and analysis. Amira Software allows you to build networks, enabling complex interactions between data objects. Biology is also complex, and biological interactions are complex networks. Complex questions require complex tools to address them. In this respect, analysis networks such as used in Amira Software outperform most other software approaches when it comes to complex data analysis. Furthermore, I like that Amira Software is quite open concerning integration of deep learning networks and scripting (Tcl, Matlab, Python,...)."
      Stephan Handschuch, PhD
      VetCore, University of Veterinary Medicine, Vienna
      Insight from your peers
      Explore industry-leading insights and research that can support your lab workflow. Amira Software can empower your lab with a cutting-edge, comprehensive imaging data analysis toolbox.
      Helping scientists answer questions that enable breakthrough discoveries in life sciences, materials science, and industry.
      Correlative Microscopy: Using Amira Software to Understand the Spread of Cancer
      Read more
      Amira Software Accelerates Veterinary Research with 3D Visualization and Analysis
      Read more
      Molecular Clarity—Discovering What’s Possible with Cryo-Electron Tomography
      Read more
      Cryo-Electron Tomography and 3D Software Advances Coronavirus Research
      Read more
      Register now to get exclusive content delivered directly to your inbox.
      Register
      Techniques and tools within Amira Software for anatomical biology
      Co-registration
      One of the main reasons researchers are using preclinical imaging is in vivo distribution studies aimed at localizing therapeutic agents in drug or gene therapy. These agents are often tagged or made susceptible for tagging with radioactive nuclei for detection with PET or SPECT imaging instruments.
      With its extensive, performance-optimized registration tools and its multi-volume architecture, Amira Software allows preclinical researchers to automatically co-register image data from any type of preclinical imaging instrumentation. Amira Software’s extensive toolbox of segmentation and quantification tools then allows for segmentation of the anatomical images and quantifying radiotracer uptake in the co-registered functional images.
      Bone separation
      Micro-CT image acquisition is one of the most popular imaging techniques for any type of bone research. One of the first steps for any bone analysis is bone segmentation and separation into individual bones.
      Amira Software offers the tools to perform bone segmentation from the surrounding tissue. The segmented bones can then be separated with Amira Software’s advanced algorithms. The individual bones can then be measured using a customizable list of the many measurements that Amira Software provides, such as size, shape and orientation.
      Cell detection
      Cellular spheroids are 3D cell cultures that represent in vivo tissue better than traditional cellular monolayers, improving the ability of researchers to study cancer, diabetes mellitus, or stem cells among many applications.
      However, cell analysis in such 3D multicellular objects is not accurately possible, especially without destroying the object of study. Amira Software can automatically separate the cells and perform volumetric, morphometric, and intensity measurements on individual cells and their nuclei in 3D image data of these spheroids. Amira Software also helps researchers with the analysis of conventional 2D cellular monolayers.
      Correlative imaging
      With its long-standing applications in multi-modality imaging techniques, Amira Software is the tool of choice for many researchers working with data from different imaging modalities. Amira Software’s architecture supports multi-volume data processing and visualization from the ground up.
      The automatic registration tools integrated into the standard edition of Amira Software with the Multi-Planar Workroom offer the flexibility of the most commonly used automated registration metrics and optimizers. These allow for the adjustment of the registration parameters to meet the needs of your data.
      Membrane detection
      Amira Software accelerates segmentation by employing automated detection of cellular features, such as filaments and membranes.
      By performing advanced image analysis and statistics on the cryo-tomograms, Amira Software combines data from these individual structures, averaging out noise and increasing contrast to create a composite model with higher resolution.
      Co-registration
      One of the main reasons researchers are using preclinical imaging is in vivo distribution studies aimed at localizing therapeutic agents in drug or gene therapy. These agents are often tagged or made susceptible for tagging with radioactive nuclei for detection with PET or SPECT imaging instruments.
      With its extensive, performance-optimized registration tools and its multi-volume architecture, Amira Software allows preclinical researchers to automatically co-register image data from any type of preclinical imaging instrumentation. Amira Software’s extensive toolbox of segmentation and quantification tools then allows for segmentation of the anatomical images and quantifying radiotracer uptake in the co-registered functional images.
      Bone separation
      Micro-CT image acquisition is one of the most popular imaging techniques for any type of bone research. One of the first steps for any bone analysis is bone segmentation and separation into individual bones.
      Amira Software offers the tools to perform bone segmentation from the surrounding tissue. The segmented bones can then be separated with Amira Software’s advanced algorithms. The individual bones can then be measured using a customizable list of the many measurements that Amira Software provides, such as size, shape and orientation.
      Cell detection
      Cellular spheroids are 3D cell cultures that represent in vivo tissue better than traditional cellular monolayers, improving the ability of researchers to study cancer, diabetes mellitus, or stem cells among many applications.
      However, cell analysis in such 3D multicellular objects is not accurately possible, especially without destroying the object of study. Amira Software can automatically separate the cells and perform volumetric, morphometric, and intensity measurements on individual cells and their nuclei in 3D image data of these spheroids. Amira Software also helps researchers with the analysis of conventional 2D cellular monolayers.
      Correlative imaging
      With its long-standing applications in multi-modality imaging techniques, Amira Software is the tool of choice for many researchers working with data from different imaging modalities. Amira Software’s architecture supports multi-volume data processing and visualization from the ground up.
      The automatic registration tools integrated into the standard edition of Amira Software with the Multi-Planar Workroom offer the flexibility of the most commonly used automated registration metrics and optimizers. These allow for the adjustment of the registration parameters to meet the needs of your data.
      Membrane detection
      Amira Software accelerates segmentation by employing automated detection of cellular features, such as filaments and membranes.
      By performing advanced image analysis and statistics on the cryo-tomograms, Amira Software combines data from these individual structures, averaging out noise and increasing contrast to create a composite model with higher resolution.
      Services
      Amira-Avizo-Introductory-training_1160x600
      Introductory training
      Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
      The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
      Amira-Avizo-advanced-training_1160x600
      Advanced training
      Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
      The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
      Amira-Avizo-Custom-Dev_1160x600
      Custom development
      With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
      We can customize and expand our software solutions at various levels.
      Resources
      Video Player is loading.
      Current Time 0:00
      Duration 0:36
      Loaded: 5.71%
      Stream Type LIVE
      Remaining Time 0:36
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Understanding the passage of air through the upper airways is crucial for planning corrective rhinosurgery when undisturbed airflow is inhibited by persistent obstructions or deformities.
        Video Player is loading.
        Current Time 0:00
        Duration 1:06
        Loaded: 14.97%
        Stream Type LIVE
        Remaining Time 1:06
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Micro-CT image acquisition is one of the most popular imaging techniques for any type of bone research. One of the first steps for any bone analysis is bone segmentation and separation into individual bones.
          Video Player is loading.
          Current Time 0:00
          Duration 1:20
          Loaded: 12.38%
          Stream Type LIVE
          Remaining Time 1:20
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Discover how Amira-Avizo Software enables the whole 3D printing workflow, from 2D image stack import, to 3D reconstruction and 3D meshing, right through to 3D printer compatible formats export. Data courtesy of National University of Singapore.
            Video Player is loading.
            Current Time 0:00
            Duration 1:04
            Loaded: 15.54%
            Stream Type LIVE
            Remaining Time 1:04
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • default, selected
              Amira Software can automatically count the cell nuclei which correspond to the cell count of the 3D spheroid cell cultures.
              Video Player is loading.
              Current Time 0:00
              Duration 1:19
              Loaded: 0.00%
              Stream Type LIVE
              Remaining Time 1:19
              1x
                • Chapters
                • descriptions off, selected
                • en (Main), selected
                Discover the capabilities of Amira Software. Transforming image data into interactive 3D models, aiding anatomical biology. Navigate high-resolution imaging from MRI, CT, light microscopy, and electron microscopy.
                Amira-Software-bone-separation_1160x600_image-card
                Courtesy of National Laboratory Animal Center - National Applied Research Laboratories, Taiwan.
                Amira-Software-coregistration_1160x600_image-card
                Co-registration for distribution studies. CT/SPECT data courtesy of Dr. Nabil Boutagy of the Yale Translation Research Imaging Center.
                Amira-Software-Preclinical_1160x600_image-card
                Maximum intensity projection and surface reconstruction of a mouse thorax. Acquisition device: VAMP TomoScope® 30s+, Amira Software - Courtesy of Dr. Elizatea Stepina and Dr. Peter Hauff, Bayer Healthcare.
                Video Player is loading.
                Current Time 0:00
                Duration 0:36
                Loaded: 9.60%
                Stream Type LIVE
                Remaining Time 0:36
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • default, selected
                  Understanding the passage of air through the upper airways is crucial for planning corrective rhinosurgery when undisturbed airflow is inhibited by persistent obstructions or deformities.
                  Video Player is loading.
                  Current Time 0:00
                  Duration 1:06
                  Loaded: 14.97%
                  Stream Type LIVE
                  Remaining Time 1:06
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • en (Main), selected
                    Micro-CT image acquisition is one of the most popular imaging techniques for any type of bone research. One of the first steps for any bone analysis is bone segmentation and separation into individual bones.
                    Video Player is loading.
                    Current Time 0:00
                    Duration 1:20
                    Loaded: 9.91%
                    Stream Type LIVE
                    Remaining Time 1:20
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      Discover how Amira-Avizo Software enables the whole 3D printing workflow, from 2D image stack import, to 3D reconstruction and 3D meshing, right through to 3D printer compatible formats export. Data courtesy of National University of Singapore.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 1:04
                      Loaded: 15.54%
                      Stream Type LIVE
                      Remaining Time 1:04
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • default, selected
                        Amira Software can automatically count the cell nuclei which correspond to the cell count of the 3D spheroid cell cultures.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 1:19
                        Loaded: 0.00%
                        Stream Type LIVE
                        Remaining Time 1:19
                        1x
                          • Chapters
                          • descriptions off, selected
                          • en (Main), selected
                          Discover the capabilities of Amira Software. Transforming image data into interactive 3D models, aiding anatomical biology. Navigate high-resolution imaging from MRI, CT, light microscopy, and electron microscopy.
                          Amira-Software-bone-separation_1160x600_image-card
                          Courtesy of National Laboratory Animal Center - National Applied Research Laboratories, Taiwan.
                          Amira-Software-coregistration_1160x600_image-card
                          Co-registration for distribution studies. CT/SPECT data courtesy of Dr. Nabil Boutagy of the Yale Translation Research Imaging Center.
                          Amira-Software-Preclinical_1160x600_image-card
                          Maximum intensity projection and surface reconstruction of a mouse thorax. Acquisition device: VAMP TomoScope® 30s+, Amira Software - Courtesy of Dr. Elizatea Stepina and Dr. Peter Hauff, Bayer Healthcare.
                          Features
                          Import and process your imaging data
                            • Handle any modality, at any scale, of any size:
                            - Bio-Formats
                            - Bitmap formats
                            - Microscopy: electron and optical
                            - Medical and neuroimage formats
                            - Molecular formats
                            - Other acquisition devices (MRI, radiography, etc.)
                            • Finite element modeling, geometric modeling, CAD
                            • Support for multi-data/multi-view, multi-channel, time series, very large data
                            • Scaling, calibration, conversion, re-sampling
                            • Image enhancement, comprehensive filtering and convolution, Fourier frequency transforms
                            • Artifact reduction algorithms
                            • Advanced multi-mode 2D/3D automatic registration
                            • Image stack alignment, arithmetic, correlation, fusion
                            Easily segment your imaging data
                            • Thresholding and auto-segmentation, object separation, automatic labeling
                            • Region growing, snakes, interpolation, wrapping, smoothing
                            • Morphological processing, including watershed and basins
                            • Machine Learning-based segmentation
                            • Automatic tracing of individual fibers and filaments
                            • Skeletonization and filament network extraction
                            • Interactive tools for generation or editing of segmentation and spatial graphs
                            • 3D surface reconstruction
                            • Grid generation for FEA/CFD
                            Export your analysis and visualization work to seamlessly publish and present it
                            • Animation and video generation
                            • Advanced key frame and object animation
                            • Mix images, geometric models, measurements and simulations
                            • Annotations, measures legends, histograms and curve plots
                            • Export spreadsheets, 3D models, high-quality images
                            • Active and passive 3D stereo vision
                            • Single and tiled screen display
                            • Immersive environment
                            Visualize and explore your imaging data
                            • Interactive high-quality volume and multichannel visualization
                            • Orthogonal, oblique, cylindrical and curved slicing
                            • Contouring and iso-surface extraction
                            • Maximum Intensity or other types of projections
                            • Vector and tensor visualization
                            • Objects and tracks
                            • Molecular visualization
                            Analyze your imaging data and obtain quantitative information
                            • Intuitive recipe creation, customization, automated replay
                            • Built-in measurements, including counts, volumes, areas, perimeters, aspect ratios and orientations
                            • User-defined measures
                            • Results viewer with spreadsheet tool and charting
                            • Automatic individual feature measurements, 3D localization and spreadsheet selection
                            • Automated statistics, distribution graphs
                            • Feature filtering using any measurement criterion
                            • Data registration, deformation, comparison and measurements
                            Easily and quickly adapt Amira Software to your specific needs
                            • Custom C++ modules development
                            • MATLAB™ bridge
                            • Python scripting API
                            Contact us
                            For Research Use Only. Not for use in diagnostic procedures.
                            Style Sheet for Global Design System
                            Style Sheet for Komodo Tabs
                            Style Sheet for Support and Service footer
                            Style Sheet for Fonts
                            Style Sheet for Cards