Search Thermo Fisher Scientific
Ion chromatography (IC) is an analytical chemistry technique that is used across the environmental, food and beverage, industrial, and pharma/biopharma industries to analyze ions and polar molecules that cannot be easily separated using high-performance liquid chromatography (HPLC). It separates and detects ions based on their interactions with a stationary phase (resin) and a mobile phase (eluent). Much like HPLC, ion chromatography systems include an autosampler, pump, separation column, and detector. In addition, IC systems include a unique component called a suppressor, which helps the peaks from sample components stand out from background noise in the chromatogram.
As a leader in IC innovation, we are committed to providing you with the latest resources to meet the ever-increasing requirements of productivity, performance and simplicity. Discover the latest technology innovations, common applications, and support resources in our IC learning center.
Learn about IC capabilities and techniques that help simplify your analyses and expand your application horizons.
For Research Use Only. Not for use in diagnostic procedures.