MDi Particle Innovation for Bioseparations
MDi technology
Thermo Fisher Scientific MDi technology combines advanced monodisperse particle platforms with bioinert hardware. MDi technology can be found in our Thermo Scientific SurePac Bio SEC MDi columns, and our Thermo Scientific ProPac 3R SAX and SCX columns.
SurePac Bio 550 SEC MDi columns - the 3 µm monodisperse silica provides superior resolution of adeno associated virus (AAV) and aggregates in cell and gene therapy applications.
ProPac 3R SAX and SCX columns - our proprietary 3 µm monodisperse polymeric technology which supports the delivery of reproducible, high resolution and robust strong anion and strong cation exchange chromatography separations for proteins, monoclonal antibodies, AAV, and other emerging modalities.
Left - 3 µm MDi polymeric monodisperse technology. Right – traditional polydisperse media Left - 3 µm MDi polymeric monodisperse technology. Right – traditional polydisperse media
Monodisperse particles
The advantages of monodisperse particles in chromatography column technology
In chromatography column technology, the choice of particle size and distribution is crucial in achieving optimal separation efficiency and reliable results.  Monodisperse particles deliver reproducibility, efficiency, enhanced resolution, and throughput, making them a superior choice in many chromatography applications.
  • Reproducibility: Offering consistent size and uniformity, our proprietary MDi monodisperse particles ensure consistent particle size distribution for column-to-column and batch-to-batch reproducibility, offering consistent results and robust, easy-to-validate methods.
  • Enhanced efficiency and resolution: Leveraging the power of monodisperse particles, the homogeneity of the packing is improved thereby reducing peak broadening effects such as eddy-diffusion. This is further improved by the small 3 µm particle size, leading to sharper and better-resolved peaks.
  • Higher throughput: 3 µm particles can be used at higher linear velocity compared to traditional 5 µm technologies, without compromising on efficiency. Moreover, the fast mass transfer rate of non-porous particles unlocks significant throughput improvements. This translates to shorter run-times and the ability to process more samples in a shorter timeframe.
Inert column hardware
Inert hardware refers to the use of materials that eliminate interactions with the sample, ensuring sample integrity and recovery. This type of hardware is particularly advantageous in biopharmaceutical analysis, where the accuracy and reliability of results are crucial. By reducing unwanted interactions, inert hardware helps to maintain sample integrity, prevent degradation or adsorption, and promote injection-to-injection reproducibility.
  • Maintains the integrity of the target molecule by minimizing interactions between the column hardware and the sample. 
  • Promotes reproducibility and consistency in analysis by eliminating or reducing interactions between the hardware and the sample.
  • Offers excellent chemical compatibility, allowing for a wide range of solvents and mobile phases to be used without compromising the integrity of the column.
  • Improves reproducibility by maintaining consistent separation performance and sample recovery across multiple runs.
ProPac 3R columns
ProPac 3R column
ProPac 3R SCX and SAX 3 μm columns provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and biosimilars.
Designed with bioinert materials to reduce secondary interactions, our ProPac 3R SCX and SAX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.
ProPac 3R SCX columns
ProPac 3R SAX columns
SurePac Bio 550 SEC MDi columns
ProPac 3R column
SurePac Bio 550 SEC MDi columns are exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process.
Designed with bioinert materials to reduce secondary interactions, SurePac Bio 550 SEC MDi columns are ideal for AAV gene therapeutics analysis and offers excellent selectivity due to the dedicated smaller particle (3 µm) size combined with wide-pore (550Å) for separating monomeric AAVs from aggregates based on their size differences.
SurePac Bio 550 SEC MDi columns
Resources
Size-exclusion chromatography of adeno-associated viruses with the SurePac Bio 550 SEC MDi column
Robust. Reproducible. Resolution. Every time. ProPac 3R IEX columns
Salt gradient analysis of Protein G using a 3 μm monodisperse SAX chromatography column
Salt gradient analysis of monoclonal antibodies using a 3 μm monodisperse SCX chromatography column
Salt gradient separation and analysis of adeno-associated virus samples using a 3 μm monodisperse strong anion exchange chromatography column
Method development for pH gradient analysis of monoclonal antibodies using a 3 μm monodisperse particle strong cation exchange chromatography column
For Research Use Only. Not for use in diagnostic procedures.