Search Thermo Fisher Scientific
We offer a comprehensive range of gas detection technologies designed to monitor oxygen, toxic and combustible gases found throughout the petrochemical, power generation, mineral processing, metal smelting and manufacturing industries, as well as in water/wastewater treatment.
When gas leaks present a threat to industrial plants, their employees and people living nearby, these detectors have the flexibility and reliability to help you to make the best decisions at a glance.
*Please note these products are available in Australia only.
Infrared point and open-path hydrocarbon gas detectors certified for use in potentially explosive atmospheres.
Searchpoint
|
Searchline Excel Edge/Excel Plus |
Searchline
|
XNX Universal
|
OELD (Optima Excel Local Display) |
---|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
![]() |
Single point gas detector | Open path gas detector | Open path gas detector | Transmitter/display to suit Optima & Excel | Display to suit Optima, Excel & Excel Edge/Plus |
HART / 4-20mA | HART / 4-20mA / Modbus / Bluetooth | 4-20mA / Modbus | HART / 4-20mA / Modbus | Bluetooth |
No indication | Status indication by means of integral LED ring | No indication | Local display with status indication | Local display with status indication |
Access via SHC1 Interrogator or XNX or OELD | Access via Smartphone/tablet | Access via SHC1 Interrogator or XNX or OELD | Access via magnetic wand | Access via Smartphone/tablet |
Optional XNX or OELD for local display | Optional OELD for local display | Optional XNX (with HART) or OELD for local display |
Transmitters providing comprehensive monitoring of combustible, toxic and Oxygen gas hazards in potentially explosive atmospheres - indoors and outdoors.
Series 3000 Transmitter |
XNX Universal
|
Sensepoint XCD Gas Detector |
Sensepoint XCD RFD Remote Flammable Gas Detector |
Sensepoint XCD RTD Remote Toxic Gas Detector |
OELD (Optima Excel Local Display) |
---|---|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
![]() |
|
Suitable for toxic or Oxygen gas sensor | Suitable for flammable, toxic or Oxygen gas sensor, plus Searchpoint Optima Plus and Searchline Excel | Suitable for flammable, toxic or Oxygen gas sensor | Suitable for flammable gas sensor (remote) | Suitable for toxic gas sensor (remote) | Suitable for Searchline Excel, Excel Edge/Plus and Searchpoint Optima Plus |
2-wire, loop powered | 3 or 4-wire, DC powered | 3-wire, DC powered | 3-wire, DC powered | 3-wire, DC powered | DC powered |
4-20mA | HART / 4-20mA / Modbus | 4-20mA / Modbus | 4-20mA / Modbus | 4-20mA / Modbus | Bluetooth |
Access via magnetic wand | Access via magnetic wand | Access via magnetic wand | Access via magnetic wand | Access via magnetic wand | Access via Smartphone/tablet |
A range of flammable, toxic and Oxygen gas sensors offering users a high quality, low cost solution to their industrial gas monitoring needs. Installation in potentially explosive atmospheres.
Series 3000/XNX Sensor Cartridges
|
Series 3000/XNX Sensor Cell |
Sensepoint
|
Sensepoint Sensor with Junction Box
|
Sensepoint XCD Sensor |
---|---|---|---|---|
![]() |
![]() |
![]() |
![]() |
![]() |
Suitable for toxic or Oxygen gas sensing | Suitable for toxic or Oxygen gas sensing | Suitable for flammable, toxic or Oxygen gas sensing | Suitable for flammable, toxic or Oxygen gas sensing | Suitable for flammable, toxic or Oxygen gas sensing |
For use with a Honeywell Series 3000 or XNX Transmitter | Replacement sensor cell for use with Series 3000 or XNX Smart Sensor Cartridge | For use with a wide range of transmitters available from Honeywell or 3rd Party | For use with a wide range of transmitters available from Honeywell or 3rd Party | For use with a Honeywell XCD Transmitter |
Smart Sensor cartridge, including replaceable sensor cell. Can be hot-swapped without powering off transmitter | Sensepoint sensor supplied with a pre-wired IP65 junction box |
Robust and reliable gas leak detector that will efficiently "hear" the loss of containment of any high-pressure gas within its detection zone.
An advanced, hazardous area ultrasonic gas leak detector, the Searchzone Sonik responds to the ultrasonic sound pressure level produced by pressurized gas leaks and is therefore unaffected by environmental conditions.
Fixed point, gas leak detector, with smartphone interface, designed to meet the needs of commercial and light industrial applications.
Detectors and controllers that use advanced technologies for detecting hundreds of gases, including hydrides, trifluorides and other semiconductor gases, detecting down to parts per billion levels.
SPM Flex Gas Detector
|
MIDAS Gas Detector |
SPM Flex Cartridges
|
MIDAS Sensor Cartridges
|
---|---|---|---|
![]() |
![]() |
![]() |
![]() |
Rugged, fixed or portable extractive gas sampler providing interference-free, Chemcassette based gas detection, for ppb level measurement | Fixed, extractive gas sampler drawing a sample locally or from a remote point to a sensor cartridge inside the detector’s chassis | Smart sensor cassettes, to suit SPM Flex gas detector | Smart sensor cartridges, to suit MIDAS gas detector |
Suitable for low level toxic gas sensing | Suitable for toxic, flammable, pyrophoric, corrosive and oxidizer (including Oxygen) gas sensing | Plug-and-play sensor cassette, with built-in RFID holding gas calibrations, alarm levels and flow settings | Plug-and-play sensor cartridges, pre-calibrated, for quick and easy replacement |
Battery or mains powered | 24V DC or PoE | Detects gas classes including Hydrides, Mineral acids, oxidisers (not oxygen), amines | Detects gas classes including Hydrides, Mineral acids, oxidisers (including oxygen), amines, LEL |
Sensitive detection using replaceable smart sensor cassettes | Sensitive detection using replaceable smart sensor cartridges |
Controllers to provide a reliable safety solution for small scale gas detection systems across a wide range of applications and industries.
Touchpoint Plus Controller |
Touchpoint Pro Controller System |
Unipoint Controller |
---|---|---|
![]() |
![]() |
![]() |
Advanced controller for up to 16 gas detection channels |
Complete centralised gas safety system, cabinet or rack mounted, monitoring up to 64 detection channels | Low-cost, single channel gas detection controller for use in indoor safe areas |
Choice of AC or DC supply voltages, with backup battery; numerous output options - 4-20mA, relay, Modbus | Options of 24Vdc powered with battery backup/UPS; numerous inputs, outputs and relay modules, Modbus, Ethernet, web-server interface | 24 VDC powered with 4-20mA and relay outputs |
7’’ colour LCD touchscreen, graphic user interface | Colour LCD touchscreen | Basic LCD display |