Beer Testing Information
Beer should ONLY be beer
Beer is the most widely consumed alcoholic beverage in the world.  Beer is typically brewed from three basic ingredients: water, a starch source brewer’s yeast, and a flavoring agent such as hops. Many varieties of beer result from differences in these ingredients, the additives used, and the brewing process. Thermo Scientific instruments help in testing the quality, consistency and purity of the final product.  
Read the blog
On-demand webinar: Automated Malt Analysis using Discrete Analyzers
Learn about new automated methods in malt analysis, with Aaron McLeod of Hartwick College, Center for Craft Food and Beverage, USA. Discrete analyzer technology offers faster, reproducible results with less sample and reagent use. We will explain experiences compared to the standard methods and will show repeatability and method comparison results.
Solutions for the Complete Brewing Process
Isohumulones form approximately 80% of the typical bitterness of beer. Their antimicrobial effect leads to a sterile beverage, their tensioactive character stabilizes the foam, and they have a major influence on the general flavor, smell, and smoothness of beer.
Quality Control & Testing for Crafting your Best Brews
As a brewer, you use both art and science to serve your customers safe, fresh beers with great character. Your business depends on quality, consistency, and flavor – which requires precision, efficiency and expertise. Learn how Thermo Scientific can support your quality testing with an unmatched combination of durable equipment, purchasing convenience and solid end-to-end customer service.
Streamlining the Test Process in a German Beer Production Facility
Radeberger Group uses only the highest quality ingredients, such as, hops and malt, allowing them to maintain their standards and produce the best beers. In fact, they have received numerous awards that provide evidence of the excellence and consistency of their brands.
Quality, Consistency, Efficiency – Helping Microbreweries Streamline Their Process
In 1887 Murphy and Son Limited was established by Albert John Murphy as a supplier of brewing components. While developing his products, Murphy realized the importance of formulating water treatment techniques that could be used to complement the specific characteristics of a beer. 
The specific aroma of beer is predominantly controlled by ingredients derived from hop, in particular the exact pattern of isohumulones. The BeerNHop solution is based on HPLC and provides an easy tool to accurately determine the isohumulone profile in the final product and in raw material for a systematic approach of innovative flavors and qualities that can make the difference. 
Quality attributes of beverage testing by integrated wet chemical analyzer
Achieving product consistency for malt, beer, wine, cider and other beverage analysis requires analysis of multiple parameters using several automated instrumentations and/or bench-top chemistry procedures. This webinar discusses how a integrated discrete analyzer simultaneously automates analysis from a single sample offering fast turnaround, reduced human error, reduced reagent use, and expanded laboratory analytical capabilities.
Search all beer testing information resources
Type  TitleCategory  Resource #
BrochureBeerCraft Software - Your Brand, Your Quality, Your Beer Molecular Spectroscopy - UV-VisBR53086
Smart NoteBeerCraft Software - How to Analyze BitternessMolecular Spectroscopy - UV-VisSN53084
Smart NoteBeerCraft Software - How to Analyze Beer ColorMolecular Spectroscopy - UV-VisSN53076
Smart NoteBeerCraft Software - How to Analyze Vicinal Diketones (VDKs)Molecular Spectroscopy - UV-VisSN53085
Poster NoteAutomated Wort and Beer Quality Control AnalysisWater Analysis InstrumentsPN71147
BrochureComplete Solution for Isohumulones in BeerChromatographyBR70746
Application NoteGradient HPLC Method for Analysis of Beer Polyphenols, Proanthocyanidins, and Bitter Acids Using a Novel Spectro-Electro Array PlatformChromatographyAN1065
Application NoteFT-NIR Analysis of Czech Republic BeerDiscrete AnalyzerAN51702
PosterDetermination of Isoxanthohumol, Xanthohumol, Alpha and Beta Bitter Acids, and trans and cis-Iso-Alpha Acids by HPLC with UV and Electrochemical Detection: Application to Hop and Beer Analysis ChromatographyPW004
Application NotebookBeer analysis applications notebook: Solutions for the complete brewing process ChromatographyAI71324
Case StudyStreamlining the Test Process in a German Beer Production FacilityDiscrete AnalyzerCS71519
Application NoteCorrelation of an Automated Discrete Analysis Sulfur Dioxide Method to Standardized Para-rosaniline Methods in the Analysis of BeerDiscrete AnalyzerAN71456
Application NoteCorrelation of the Free Amino Nitrogen and Nitrogen by O-Phthaldialdehyde Methods in the Assay of BeerDiscrete AnalyzerAN71798
Case StudyQuality, Consistency, Efficiency – Helping Microbreweries Streamline Their Process Discrete AnalyzerCS71723
Application NoteApplications Notebook: Alcoholic Beverages ChromatographyLPN3086
WebinarNovel LC Approaches to Analysis of Alcoholic Beverages - From Fast and Easy Quality Control to Exploring the SensometabolomesChromatography 
FlipbookBeer Should ONLY be Beer. Application Summary CompendiumChromatographyAB71410
Application NoteIon Chromatography: A Versatile Technique for the Analysis of BeerDiscrete AnalysisAN71410
Smart NoteWhat is Better for Automating Wet Chemical Analysis? Integrated Discrete Analyzer or Flow Analyzers?Wet Chemical AnalysisSN73521
InfographicGallery Discrete Analyzer - Increase Productivity and Profitability in Contract Testing LabsWet Chemical AnalysisIN73408
eBookAnalytical Guide for Routine Beverage AnalysisWet Chemical AnalysisEB73110
FlyerAutomate your wet chemical analysisDiscrete AnalyzerFL71241
Application BriefCitric acid determination in mixed beer beveragesAB001056
Right color? Right taste?
Be sure with UV-Vis
Ensure batch-to-batch consistency with BeerCraft Software and GENESYS 50 or 150 UV-Vis Spectrophotometers. Tests are preprogrammed for fast, easy quality analysis.
Learn more
FTIR spectrometers for quality control