What is noble gas mass spectrometry?
Noble gas mass spectrometry studies isotopes of noble gases helium (He), neon (Ne), argon (Ar), krypton (Kr), and xenon (Xe). These gases are found in very small quantities in a variety of environmental samples and the study of the isotopes of these noble gases provides powerful insights in the fields of geochronology, cosmochemistry, and thermochronology.
Isotope ratio analysis of the small samples of noble gases require static vacuum, low-volume, high-resolution instrumentation. The three Thermo Scientific noble gas mass spectrometers are designed to meet these requirements.
Whether for Ar-Ar dating, where sensitivity and multicollection is key, or for high precise He isotope analysis requiring low abundance sensitivity, or for simultaneous interference-free measurement of all noble gases, where ultimate high-resolution variable multicollection is mandatory, our Noble Gas Portfolio covers it all.
We also offer a noble gas preparation system, to provide a source of clean noble gases (helium, neon, argon, krypton, and xenon) for analysis.
Improve your geochronology analysis
Argus VI Static Vacuum Mass Spectrometer
Improve the precision of Ar-Ar dating by an order of magnitude. Determine the isotope ratios of all five argon isotopes simultaneously with the Thermo Scientific Argus VI Static Vacuum Mass Spectrometer. 
Learn more about Argus VI Static Vacuum Mass Spectrometer ›
Explore new research areas with sub-permil precision
Helix SFT Split Flight Tube Noble Gas Mass Spectrometer
Enter new fields of research with unique sub-permil precision by using the new Thermo Scientific Helix SFT Split Flight Tube Noble Gas Mass Spectrometer.
Learn more about Helix SFT Split Flight Tube Noble Gas Mass Spectrometer ›
Achieve high-resolution and high-precision results
Helix MC Plus Multicollector Noble Gas Mass Spectrometer
Redetermine the isotope ratios of neon, argon, krypton and xenon with the outstanding high-resolution and precision of the Thermo Scientific Helix MC Plus Multicollector Noble Gas Mass Spectrometer.
Learn more about Helix MC Plus Multicollector Noble Gas Mass Spectrometer ›
Argus VI Mass Spectrometer
Helix SFT Mass Spectrometer
Helix MC Plus Mass Spectrometer
Overview
Low volume, high sensitivity for best precision for small samples.
Unique ability to measure both Helium isotopes simultaneously.
Ultra high-resolution & resolving power for wide variety of noble gas  applications.
Analyzer
Fixed collector array – incorporating advanced Faraday (1e10 to 1e13) and low noise CDD SEM technology
Beam deflection technology – to enable a variety of multicollector configurations and to cater for range of sample sizes
Fixed collector array – to minimize internal volume
ESA / SEM for ultimate abundance sensitivity
Moveable collectors - for maximum versatility and precision
Combined Faraday Multiplier (x5) - for wide dynamic range
Mass range
A multicollector static vacuum mass spectrometer capable of measuring any mass between 1 and 140.
A multicollector static vacuum mass spectrometer capable of measuring any mass between 1 and 140.
A multicollector static vacuum mass spectrometer capable of measuring any mass between 1 and 140.
Key technology
Lowest commercially available internal volume Emission Suppression Technology
Split flight tube allows isotopes of helium to be measured simultaneously
An instrument with the ability to resolve every major interference from every noble gas isotope
Key applications
Ar-Ar dating of geological samples
Groundwater dating, low temperature thermochronology, composition of Earth’s mantle.
Understanding origin and evolution of the Solar system.
81Kr analysis for groundwater dating.
Xe isotopic ratios in meteorites to studying the evolution of the Solar system.
Qtegra ISDS Software for noble gas MS
Discover the Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS) Software platform, and experience a new level of flexibility, productivity and ease of use for your noble gas analyses.
Learn more about Qtegra ISDS Software ›
Amplify your precision with 1013 amplifier technology
Are you limited in sample size? One of the major challenges in geosciences is the analysis of small ion beams. The analytical precision of these studies is limited by the detection system of your mass spectrometer. Equipped with 1013 ohm amplifiers with a new resistor design, our spectrometers enable you to get faster response times and extremely low noise/signal characteristics.
Learn more about 1013 Ohm Technology ›
Analysis without compromise
The patented Emission Suppression Technology (EST) revolutionizes noble gas isotope ratio mass spectrometry by switching off ionization whilst the sample is equilibrating in the source. This removes the trade-off between sample equilibration and precision.
Learn more about EST ›
Applications
1013 Ω Amplifiers for the Analysis of Noble Gases
The improved signal/noise of the amplifiers equipped with a 1013 Ω feedback resistor represents a decisive improvement in small sample analysis.
  Download tech note ›
Argon Isotope Ratio Measurements
Improve the precision of Ar-Ar dating by an order of magnitude. Determine the isotope ratios of all five argon isotopes simultaneously with the Argus VI SVMS.
  Download app note ›
Argon-Argon Dating
The Argus VI SVMS performs noble gas analysis with beam deflection and 1013 Ohm Amplifier technology, reducing system volume.
* Required field
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes