Spot On Faster Path to MS Analysis
Mass spec ionization that is on the faster path to MS analysis
If you are dealing with a rising backlog of samples due to an insufficient chromatography-based solution, the Thermo Scientific VeriSpray PaperSpray ion source for mass spectrometry is a valuable alternative solution for MS analysis. This paper spray ionization technique provides direct sample analysis capability with your triple quadrupole mass spectrometer, eliminating the need for chromatographic based separation, simplifying your workflow, getting to results faster, and reducing cost per test.
VeriSpray PaperSpray ion source for mass spectrometry
On this page
Watch video on how VeriSpray ion source works
Learn about how the VeriSpray ion source for mass spectrometry enables quick and cost-effective triple quadrupole mass spectrometry analysis without the hassle of complex chromatographic-based separations.
Key benefits of VeriSpray paper spray ionization technique
Reduce sample turnaround prior to MS analysisLower costs per testSeamless data acquisitions and processing
Run up to 240 samples unattended for high throughput analysis of unprocessed samples. MS results can be generated in less than 2 minutes due to minimal sample preparation and the elimination of chromatography using the ion source.Bring more MS based tests in house with the easy-to-use VeriSpray ion source that does not require any chromatography user background. Decrease your solvent consumption and bid farewell to instrument maintenance costs.VeriSpray ion source can be directly controlled by the TSQ Series II ICSW 3.2 Tune editor which includes method templates for workflows. Utilize the Thermo Scientific Xcalibur 4.2 SP1 for easy and efficient data processing.
A remarkably simplified workflow with VeriSpray ion source
Direct MS-based ionization of complex sample matrices spotted on a paper using the VeriSpray ion source instead of time-consuming sample preparation and chromatography. VeriSpray paper spray ionization technology is a fast, easy sample introduction technique that uses a direct sampling process that introduce samples to the mass spectrometer via electrospray ionization. It simply adds sample and solvent to the paper cartridge and loads it into the system.
VeriSpray ion source for mass spectrometry lowers costs per test
The VeriSpray ion source allows use of “dilute and shoot” methods to minimize solvent consumption and time-consuming sample preparation steps like derivatization. By making the specificity and sensitivity of mass spectrometry results available to non-experts, you can run more tests in your analytical testing lab, saving money spent on sample send-outs.
Fast quantification of antifungal drugs with the VeriSpray ion source coupled to Thermo Scientific TSQ Altis mass spectrometer.
Quantification of antifungal drugs with the VeriSpray ion source coupled to the TSQ Altis mass spectrometer
For Research Use Only. Not for use in diagnostic procedures.
VeriSpray ion source ordering guide
Paper Spray Ionization: The Faster Path to Mass Spectrometry Analysis
Using paper spray ionization techniques, and the Thermo Scientific VeriSpray PaperSpray ion source, researchers in clinical and forensic toxicology labs with high throughput requirements can now perform direct mass spectrometry-based analyses with minimal solvent consumption and sample prep, thereby saving costs per test and reducing the time involved.
Download eBook
How paper spray mass spectrometry facilitated harm reduction drug checking
Download this case study to learn more about how the AERL used Thermo Scientific VeriSpray PaperSpray ion source connected to the Thermo Scientific TSQ Fortis triple quadrupole mass spectrometer to meet their needs for an on-site testing program and the impressive results they achieved.
Download Case Study
Ideal applications for using VeriSpray ion source for mass spectrometry
Related LC-MS systems