1500 Series A2 BSC
Made in USA – for North America, Latin America, APAC, and Emerging Markets
The Thermo Fisher Series 1500 Class II, Type A2 Biological Safety Cabinet offers advanced protection, energy efficiency and expectational user comfort and convenience with upgrades such as touchscreen and smart airflow technologies. This cabinet is an essential upgrade for any lab.
Enhanced protection, convenience, and comfort
Convenience
7 inch touchscreen display offers at-a-glance system health, operational status and performance data. 
bsc1500 convenience card
Containment
Dual blower system monitors and controls airflow balance of both inflow and downflow in real-time, which is critical for helping maintain cleanliness and containment.
bsc1500 containment card
Comfort
SmartClean and tactile window mechanisms are designed with user comfort in mind. An array of ergonomic accessories are available. 
bsc1500 comfort card
Features
bsc 1500 us features
  1. Advanced Airflow Control - Thermo Scientific SmartFlow Plus and DAVe (Digital Airflow Verification) Plus technology, featuring separate downflow and exhaust fans for superior airflow management. 
  2. Touchscreen convenience - Intuitive 7” touchscreen interface to support real-time alarm and performance monitoring.
  3. Tactile window mechanism - Proprietary window positioning mechanism that helps provide haptic feedback to easily locate the correct window position.
  4. Energy efficiency - When front sash is closed, blower speed is automatically reduced, offering sample protection at reduced energy consumption even when the cabinet is not in use.
  5. Contamination control - Programmable UV light reduces risk of sample contamination. Allows timed preset to run disinfection cycle prior to working. 
  6. Ease-of-cleaning - Proprietary Thermo Scientific SmartClean front window design improves user access to the work area and enables cleaning of hard to reach areas.
  7. Operator comfort - Clean and safe routing of vacuum tubing and cables through SmartPort access in the double side walls. Various ergonomic-centric accessories are available to help ensure a comfortable and efficient workspace.
Applications
All applications requiring high-level protection and enhanced user comfort with touchscreen.
Typical markets: Academia, Biotech and Pharma.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific use or application.
Certifications
  • NSF/ANSI 49, NRTL (UL), CE, CULus
Resources
Videos
Ordering information
Available in: North America, Latin America, APAC, and Emerging Markets
Cabinet width
Chamber material choices
Ordering details
3 ft, 4 ft, 6 ft Stainless steel, SmartCoat 120V
3 ft, 4 ft, 6 ft
Stainless steel, SmartCoat
230V
3 ft, 4 ft, 6 ft
Stainless steel, SmartCoat Accessories