In addition to our custom synthesis options for oligos, primers, probes, and genes, Thermo Fisher Scientific offers a selection of other high-quality, pre-designed DNA primers and oligos. These products include random primers and oligo (dT) primers for first strand cDNA synthesis, gene specific primers for sequencing, forward and reverse sequencing primers and kits, and the MegaPlex RT Primers and Megaplex primer pools developed for assessing expression profiles.
Review our pre-designed, pre-synthesized offerings that are commonly used in a variety of molecular biology and genomic applications ranging from viral detection, construction of cDNA libraries, expression analyses, RNAi-based studies, epigenetic research, next generation sequencing, and more.
Random DNA primers and hexamers
Random hexamers product photo
Random primers and hexamers are short oligodeoxyribonucleotides with random base sequences (usually [d(N)6]), typically used to prime mRNAs with or without poly(A) for cDNA synthesis. Unlike oligo(dT) primers, these primers are not template specific and are suitable for DNA synthesis using Klenow fragments with DNA templates or for cDNA synthesis using reverse transcriptase with mRNA templates.
Learn more about their application in reverse transcription reactions
Order Invitrogen random primers and hexamers
Catalog # Name Size Price (USD) Qty
N8080127 Random Hexamers (50 μM) Each
85.40
Special offer
Online exclusive
Ends: 01-Jun-2025
122.00
Save 36.60 (30%)
48190011 Random Primers Each
82.60
Special offer
Online exclusive
Ends: 01-Jun-2025
118.00
Save 35.40 (30%)
AM5722G Random Decamers (50 μM) Each
176.00
Oligo (dT) primers
Invitrogen oligo (dT) primers product photo
Typically, oligo (dT) primers are a string of 12–20 deoxythymidines that specifically anneal to poly(A) tails of eukaryotic mRNAs. In contrast to random primers, oligo (dT) primers are not suitable for degraded samples but are excellent for constructing cDNA libraries and recommended for use with reverse transcriptases (RTs) as Superscript IV RT.
Order Invitrogen oligo(dT) primers
Sequencing primers
M13 reverse primer sequencing primers product photo
Common sequences of Short DNA oligonucleotides are often used as primers for sequencing cloned genes (Sanger sequencing). Choose from our commonly used sequencing primers to initiate your sequence verification projects.
Learn about sequencing applications
Order sequencing primers and kits
Primer pools
Different colors portraying different primer pool assays
Pools of pre-designed primers can help generate comprehensive expression profiles, especially where assay sensitivity is critically important. The collection of MegaPlex RT Primers and Megaplex Primer Pools significantly enhance the ability to detect microRNAs expressed at low levels with as low as 1 ng of input total RNA.
 Read how Megaplex Primer Pools were used to profile miRNAs with limited samples
Order MegaPlex RT primers and Megaplex primer pools
Random DNA primers and hexamers
Random hexamers product photo
Random primers and hexamers are short oligodeoxyribonucleotides with random base sequences (usually [d(N)6]), typically used to prime mRNAs with or without poly(A) for cDNA synthesis. Unlike oligo(dT) primers, these primers are not template specific and are suitable for DNA synthesis using Klenow fragments with DNA templates or for cDNA synthesis using reverse transcriptase with mRNA templates.
Learn more about their application in reverse transcription reactions
Order Invitrogen random primers and hexamers
Catalog # Name Size Price (USD) Qty
N8080127 Random Hexamers (50 μM) Each
85.40
Special offer
Online exclusive
Ends: 01-Jun-2025
122.00
Save 36.60 (30%)
48190011 Random Primers Each
82.60
Special offer
Online exclusive
Ends: 01-Jun-2025
118.00
Save 35.40 (30%)
AM5722G Random Decamers (50 μM) Each
176.00
Oligo (dT) primers
Invitrogen oligo (dT) primers product photo
Typically, oligo (dT) primers are a string of 12–20 deoxythymidines that specifically anneal to poly(A) tails of eukaryotic mRNAs. In contrast to random primers, oligo (dT) primers are not suitable for degraded samples but are excellent for constructing cDNA libraries and recommended for use with reverse transcriptases (RTs) as Superscript IV RT.
Order Invitrogen oligo(dT) primers
Catalog # Name Size Price (USD) Qty
18418012 Oligo(dT)12-18 Primer Each
82.60
Special offer
Online exclusive
Ends: 01-Jun-2025
118.00
Save 35.40 (30%)
18418020 Oligo(dT)20 Primer Each
85.40
Special offer
Online exclusive
Ends: 01-Jun-2025
122.00
Save 36.60 (30%)
N8080128 Oligo d(T)16 (50 μM) Each
84.70
Special offer
Online exclusive
Ends: 01-Jun-2025
121.00
Save 36.30 (30%)
12577011 Anchored Oligo(dT)20 Primer Each
160.30
Special offer
Online exclusive
Ends: 01-Jun-2025
229.00
Save 68.70 (30%)
AM5730G Oligo (dT) Primer (50 μM) Each
165.65
Online Exclusive
176.00
Save 10.35 (6%)
AM5712 T7-Oligo(dT)24V Promoter Primer, Anchored (100 ng/μl) Each
315.00
Sequencing primers
M13 reverse primer sequencing primers product photo
Common sequences of Short DNA oligonucleotides are often used as primers for sequencing cloned genes (Sanger sequencing). Choose from our commonly used sequencing primers to initiate your sequence verification projects.
Learn about sequencing applications
Order sequencing primers and kits
Primer pools
Different colors portraying different primer pool assays
Pools of pre-designed primers can help generate comprehensive expression profiles, especially where assay sensitivity is critically important. The collection of MegaPlex RT Primers and Megaplex Primer Pools significantly enhance the ability to detect microRNAs expressed at low levels with as low as 1 ng of input total RNA.
 Read how Megaplex Primer Pools were used to profile miRNAs with limited samples
Order MegaPlex RT primers and Megaplex primer pools
Find everyday lab essentials to power your PCR every step of the way. Shop now
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.