DNA helix as cloud
The polymerase chain reaction (PCR) is a sensitive and efficient method for amplifying a single copy of a target DNA sequence to millions of copies. Target DNA detection and/or amplification by PCR is an important step in cloning, gene expression analysis, genotyping, sequencing, and mutagenesis. PCR has a broad range of applications, including in research for infectious diseases, cancer, forensic analysis, and agricultural biotechnology.
For efficient endpoint PCR with fast and reliable results, here are five key steps to consider:
DNA isolation
DNA isolation icon on a purple background
DNA isolation is a crucial first step in the PCR workflow, whether you are isolating genomic DNA (gDNA) or bacterial plasmids. Selecting nucleic acid purification products that are optimized to provide maximum yield, purity, and integrity from virtually any sample type and application is important for your research success.
Common nucleic acid isolation methods include:
 Organic extraction
Tube with three layers of liquids
Spin column
Tube with column inside
Magnetic beads
Tube with magnetic beads inside and magnet outside
Benefits
  • Efficient lysis of cells and tissue
  • Rapid denaturation of nucleases
  • Stabilization of nucleic acids
  • Ease of use
  • High yield and purity
  • Throughput flexibility
  • No risk of clogging
  • Increased target capture efficiency
  • Rapid collection and concentration of sample
  • Ability to automate
  • Scalability
Product offerings
To determine which purification kit is appropriate for your DNA type and sample type, use our interactive selection guide
Tired of purifying DNA for PCR?
direct-PCR
Check out a universal direct PCR master mix that works with different sample types. No DNA purification needed!
Primer design
Oligo icon on a red background
Good design (i.e., good sequence selection) and high-quality primers are critical to your PCR reactions. In general, a length of 18–30 nucleotides for primers is optimal. The melting temperatures of the primers should be between 65°C and 75°C, and within 5°C of each other.
Forget the hassle of designing primers whether you are handling one or 50 genes. Take advantage of the benefits offered by our robust and easy-to-use Primer3-based OligoPerfect Designer
Custom oligos manufacturing
From the start of your oligos order to follow ups and special requests, we are here to meet your needs.
Video Player is loading.
Current Time 0:00
Duration 1:41
Loaded: 4.95%
Stream Type LIVE
Remaining Time 1:41
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Ordering custom DNA oligos
    The appropriate synthesis scale and purification for your application depend on the nature of your downstream applications. We offer oligos made to your specifications, with rigorous quality control, and quick turnaround for PCR and a variety of other applications. Learn more about Invitrogen custom DNA oligos
    Oligos ordering tools
    Here's a way to share your oligo orders directly with your team, giving them the ability to manage and track shipments.
    Video Player is loading.
    Current Time 0:00
    Duration 0:59
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 0:59
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      PCR enzyme selection
      Microcentrifuge tube icon on a grey background
      DNA polymerase is an essential component for PCR because of its key role in synthesizing new DNA strands. Because of the sensitive and specific nature of PCR, it is important to choose high-quality enzymes and reagents to produce optimal results.
      A comprehensive portfolio of PCR enzymes and master mixes is available with the high performance and consistency you need. View our online selection guide to find the best enzyme for common PCR applications.
      What are benefits of a universal annealing temperature?
      Learn the importance of the annealing step in PCR, how to circumvent optimization steps using a specially formulated PCR buffer, and the benefits of universal primer annealing enabled by the buffer.
      Video Player is loading.
      Current Time 0:00
      Duration 2:40
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 2:40
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Need dNTPs for your PCR? Browse all nucleotides
        Thermal cycling
        PCR consists of heating and cooling cycles (thermal cycling) for DNA amplification. Therefore, the success or failure of a PCR reaction may depend on selecting the right thermal cycler (which automates the process) and PCR plastics (which holds the reaction).
        Thermal cycler background on a dark blue background
        Applied Biosystems thermal cyclers enable precise, consistent results for every challenge, application, and budget. Since 1987, our instruments have built a reputation for reliability, temperature accuracy, user-friendly interfaces, and innovation.
        Deciding on a thermal cycler for your lab? Discover our offerings for one that fits your need.
        ProFlex PCR system
        ProFlex PCR System fits how you work today and tomorrow with 5 interchangeable block formats – maximize your throughput or run three experiments at once.
        Video Player is loading.
        Current Time 0:00
        Duration 1:16
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 1:16
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          PCR plastics
          Plate icon on a grey background
          Using the right PCR plastics for your application and instrument can improve the reliability of your PCR results. Regardless of the plastics format you select, proper fit and uniform heat transfer during thermal cycling are essential for successful PCR.
          Applied Biosystems PCR plastics have been designed and validated to work with Applied Biosystems thermal cyclers for more than 25 years. To find plastics compatible with Applied Biosystems and other thermal cyclers, use our PCR Plastics Selection Tool
          How to choose a PCR plate
          Find the right plastics based on format for your PCR and qPCR instrument.
          Video Player is loading.
          Current Time 0:00
          Duration 2:49
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 2:49
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            PCR plastics offerings
            Amplicon analysis
            Icon of DNA gel bands on a purple background
            Nucleic acid electrophoresis is a common technique in the PCR workflow to separate, identify, quantify, and/or purify amplicons. Choosing the right tools for nucleic acid electrophoresis can significantly improve and accelerate results, enabling you to address downstream applications sooner.
            Invitrogen E-Gel precast agarose gel system offers fast and sensitive analysis of PCR amplicons while minimizing time-consuming and messy prep work. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Analysis of PCR products can be completed in as little as 15 minutes.
            E-Gel_running_time
            Invitrogen E-Gel Power Snap System
            The E-Gel Power Snap System combines rapid, real-time nucleic acid analysis with high-resolution image capture to reduce workflow time and accelerate discovery.
            Video Player is loading.
            Current Time 0:00
            Duration 1:16
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 1:16
            1x
              • Chapters
              • descriptions off, selected
              • en (Main), selected
              Recommended products for nucleic acid electrophoresis
              Pouring your own agarose gels? Explore reagents optimized for success
              Helpful links for nucleic acid electrophoresis
              DNA isolation
              DNA isolation icon on a purple background
              DNA isolation is a crucial first step in the PCR workflow, whether you are isolating genomic DNA (gDNA) or bacterial plasmids. Selecting nucleic acid purification products that are optimized to provide maximum yield, purity, and integrity from virtually any sample type and application is important for your research success.
              Common nucleic acid isolation methods include:
               Organic extraction
              Tube with three layers of liquids
              Spin column
              Tube with column inside
              Magnetic beads
              Tube with magnetic beads inside and magnet outside
              Benefits
              • Efficient lysis of cells and tissue
              • Rapid denaturation of nucleases
              • Stabilization of nucleic acids
              • Ease of use
              • High yield and purity
              • Throughput flexibility
              • No risk of clogging
              • Increased target capture efficiency
              • Rapid collection and concentration of sample
              • Ability to automate
              • Scalability
              Product offerings
              To determine which purification kit is appropriate for your DNA type and sample type, use our interactive selection guide
              Tired of purifying DNA for PCR?
              direct-PCR
              Check out a universal direct PCR master mix that works with different sample types. No DNA purification needed!
              Primer design
              Oligo icon on a red background
              Good design (i.e., good sequence selection) and high-quality primers are critical to your PCR reactions. In general, a length of 18–30 nucleotides for primers is optimal. The melting temperatures of the primers should be between 65°C and 75°C, and within 5°C of each other.
              Forget the hassle of designing primers whether you are handling one or 50 genes. Take advantage of the benefits offered by our robust and easy-to-use Primer3-based OligoPerfect Designer
              Custom oligos manufacturing
              From the start of your oligos order to follow ups and special requests, we are here to meet your needs.
              Video Player is loading.
              Current Time 0:00
              Duration 1:41
              Loaded: 2.77%
              Stream Type LIVE
              Remaining Time 1:41
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Ordering custom DNA oligos
                The appropriate synthesis scale and purification for your application depend on the nature of your downstream applications. We offer oligos made to your specifications, with rigorous quality control, and quick turnaround for PCR and a variety of other applications. Learn more about Invitrogen custom DNA oligos
                Oligos ordering tools
                Here's a way to share your oligo orders directly with your team, giving them the ability to manage and track shipments.
                Video Player is loading.
                Current Time 0:00
                Duration 0:59
                Loaded: 0.00%
                Stream Type LIVE
                Remaining Time 0:59
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  PCR enzyme selection
                  Microcentrifuge tube icon on a grey background
                  DNA polymerase is an essential component for PCR because of its key role in synthesizing new DNA strands. Because of the sensitive and specific nature of PCR, it is important to choose high-quality enzymes and reagents to produce optimal results.
                  A comprehensive portfolio of PCR enzymes and master mixes is available with the high performance and consistency you need. View our online selection guide to find the best enzyme for common PCR applications.
                  What are benefits of a universal annealing temperature?
                  Learn the importance of the annealing step in PCR, how to circumvent optimization steps using a specially formulated PCR buffer, and the benefits of universal primer annealing enabled by the buffer.
                  Video Player is loading.
                  Current Time 0:00
                  Duration 2:40
                  Loaded: 0.00%
                  Stream Type LIVE
                  Remaining Time 2:40
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • en (Main), selected
                    Need dNTPs for your PCR? Browse all nucleotides
                    Thermal cycling
                    PCR consists of heating and cooling cycles (thermal cycling) for DNA amplification. Therefore, the success or failure of a PCR reaction may depend on selecting the right thermal cycler (which automates the process) and PCR plastics (which holds the reaction).
                    Thermal cycler background on a dark blue background
                    Applied Biosystems thermal cyclers enable precise, consistent results for every challenge, application, and budget. Since 1987, our instruments have built a reputation for reliability, temperature accuracy, user-friendly interfaces, and innovation.
                    Deciding on a thermal cycler for your lab? Discover our offerings for one that fits your need.
                    ProFlex PCR system
                    ProFlex PCR System fits how you work today and tomorrow with 5 interchangeable block formats – maximize your throughput or run three experiments at once.
                    Video Player is loading.
                    Current Time 0:00
                    Duration 1:16
                    Loaded: 0.00%
                    Stream Type LIVE
                    Remaining Time 1:16
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      PCR plastics
                      Plate icon on a grey background
                      Using the right PCR plastics for your application and instrument can improve the reliability of your PCR results. Regardless of the plastics format you select, proper fit and uniform heat transfer during thermal cycling are essential for successful PCR.
                      Applied Biosystems PCR plastics have been designed and validated to work with Applied Biosystems thermal cyclers for more than 25 years. To find plastics compatible with Applied Biosystems and other thermal cyclers, use our PCR Plastics Selection Tool
                      How to choose a PCR plate
                      Find the right plastics based on format for your PCR and qPCR instrument.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 2:49
                      Loaded: 0.00%
                      Stream Type LIVE
                      Remaining Time 2:49
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • en (Main), selected
                        PCR plastics offerings
                        Amplicon analysis
                        Icon of DNA gel bands on a purple background
                        Nucleic acid electrophoresis is a common technique in the PCR workflow to separate, identify, quantify, and/or purify amplicons. Choosing the right tools for nucleic acid electrophoresis can significantly improve and accelerate results, enabling you to address downstream applications sooner.
                        Invitrogen E-Gel precast agarose gel system offers fast and sensitive analysis of PCR amplicons while minimizing time-consuming and messy prep work. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Analysis of PCR products can be completed in as little as 15 minutes.
                        E-Gel_running_time
                        Invitrogen E-Gel Power Snap System
                        The E-Gel Power Snap System combines rapid, real-time nucleic acid analysis with high-resolution image capture to reduce workflow time and accelerate discovery.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 1:16
                        Loaded: 0.00%
                        Stream Type LIVE
                        Remaining Time 1:16
                        1x
                          • Chapters
                          • descriptions off, selected
                          • en (Main), selected
                          Recommended products for nucleic acid electrophoresis
                          Pouring your own agarose gels? Explore reagents optimized for success
                          Helpful links for nucleic acid electrophoresis
                          Find everyday lab essentials to power your PCR every step of the way. Shop now
                          For Research Use Only. Not for use in diagnostic procedures.