custom-barcodes-image-230x195 copy
Bring the power of barcoding to your process
Custom barcoding and labeling are available for Abgene and Nunc Storage Plates to enhance tracking of samples and groups of samples.
Custom barcoding for your plates
Custom barcoding for Nunc plates
Custom nunc barcoding
  • Choose from 5 code types to best meet your application needs
  • Select your label location–apply to one or more plate sides
  • No repeats–guaranteed
  • Human-readable option, including positions and highlight color
  • Sequences archived for data management
Request custom barcoding at:
thermofisher.com/barcodeconfigurator
Custom barcoding for Abgene plates
Custom ABgene barcoding
  • Choose from three codes: Code 128, Code 39, Interleaved 2 of 5, with flexible human-readable code position
  • Label sizes customizable according to requirements
  • Barcode density range of dimensions available
  • Choose either long side or short side for barcode
  • 1,000 plate minimum orders (smaller quantities may be possible, but are subject to an order fee)
Please contact your sales rep for details on ordering.
Nunc barcode configurator
  • Choose from 5 code types to best meet your application needs
  • Select your label location – apply to one or more plate sides
  • No repeats – guaranteed!
  • Human-readable option, including positions and highlight color
  • Sequences archived for data management
  • Print a sample barcode to ensure compatibility with your reader
Barcoding services for your cryogenic tubes
Nunc and Nalgene tubes
Cryogenic tubes
Pre-label Nunc or Nalgene Cryogenic Tubes with barcodes for easy tracking of samples in databases, LIMS or Excel files.
  • Robust codes are direct-printed onto white patches for high contrast, durability and readability
  • Available with human readable text for the ability to read without barcode readers
  • Barcodes are resistant to isopropyl alcohol, 100% butanol, 10-20% DMSO, 5% bleach, 10% acetic, hydrochloric acids and 10% NaOH
  • Virgin grade polypropylene resins reduce the risk of extractables and leachables interfering with samples
  • Strict quality management and processes in ISO 13485 manufacturing facilities
  • Tubes are guaranteed with a Sterility Assurance Level of 10-3 or 10-6 by gamma irradiation
  • No risk of inhibiting sample integrity through manufacturing by-products
  • Tubes are RNase/DNase, pyrogen, and cytotoxin free
  • We guarantee that your barcode will never be duplicated at manufacture
Choose non-custom codes or customize codes to fit any system.
Non-custom Tubes
  • Non-custom codes available for Nalgene and Nunc Cryogenic tubes for most popular sizes
Custom Tubes
  • Custom codes available for nunc or Nalgene tubes include code 128 (subset A & B or subset C), Interleave 2 of 5 and Code 39
Support
Need technical support? Contact our expert team for technical and application support of Laboratory Products