Madin-Darby canine kidney (MDCK) cells.
Viable Madin-Darby canine kidney (MDCK) cells sequentially stained with BODIPY® FL C5-ceramide (Cat. No. D3521, B22650), LysoTracker® Red DND-99 (Cat. No. L7528) and Hoechst 33258 (Cat. No. H1398, H3569, H21491). Green-fluorescent BODIPY® FL C5-ceramide localized to the Golgi apparatus, red-fluorescent LysoTracker® Red stain accumulated in the lysosomes, and the blue-fluorescent Hoechst 33258 dye stained the nuclei. The multiple-exposure image was acquired with bandpass filters appropriate for fluorescein, Texas Red® dye and DAPI.
Viable Madin-Darby canine kidney (MDCK) cells sequentially stained with BODIPY® FL C<sub>5</sub>-ceramide (Cat. No. D3521, B22650), LysoTracker® Red DND-99 (Cat. No. L7528) and Hoechst 33258 (Cat. No. H1398, H3569, H21491). Green-fluorescent BODIPY® FL C<sub>5</sub>-ceramide localized to the Golgi apparatus, red-fluorescent LysoTracker® Red stain accumulated in the lysosomes, and the blue-fluorescent Hoechst 33258 dye stained the nuclei. The multiple-exposure image was acquired with bandpass filters appropriate for fluorescein, Texas Red® dye and DAPI.
Related Products
Catalog # Name Size Price (USD) Qty
B22650 BODIPY™ FL C5-Ceramide complexed to BSA Each
438.00
H1398 Hoechst 33258, pentahydrate (bis-benzimide), 100 mg, 100 mg Each
106.65
Online Exclusive
109.00
Save 2.35 (2%)
H3569 Hoechst 33258, pentahydrate (bis-benzimide), 10 mg/mL solution in water, 10 mL Each
139.65
Online Exclusive
144.00
Save 4.35 (3%)
Related Images