Staining with Arg-CMAC.
Saccharomyces cerevisiae vacuolar lumen stained with 7-amino-4-chloromethylcoumarin, L-arginine amide (Arg-CMAC), available in our Yeast Vacuole Marker Sampler Kit (Cat. No. Y7531). This kit contains three additional fluorescent vacuolar lumen stains plus a green-fluorescent vacuolar membrane stain. After staining, yeast were viewed by epifluorescence and DIC microscopy.
<i>Saccharomyces cerevisiae</i> vacuolar lumen stained with 7-amino-4-chloromethylcoumarin, L-arginine amide (Arg-CMAC), available in our Yeast Vacuole Marker Sampler Kit (Cat. No. Y7531). This kit contains three additional fluorescent vacuolar lumen stains plus a green-fluorescent vacuolar membrane stain. After staining, yeast were viewed by epifluorescence and DIC microscopy.
Related Products
Catalog # Name Size Price (USD) Qty
Y7531 Yeast Vacuole Marker Sampler Kit 1 kit
447.00
Related Images