HeLa cells labeled with SYTO® 9
Live HeLa cells labeled with 5 µM SYTO® 9 (Cat. No. S34854) for 30 minutes at 37°C.
Live HeLa cells labeled with 5 µM SYTO® 9 (Cat. No. S34854) for 30 minutes at 37°C.
Related Products
Catalog # Name Size Price (USD) Qty
S34854 SYTO™ 9 Green Fluorescent Nucleic Acid Stain, 100 μL Each
393.65
Online Exclusive
405.00
Save 11.35 (3%)
Related Images