Multiple Myeloma Monitoring Guidelines for Relapse Detection
The International Myeloma Working Group (IMWG) produce guidelines for monitoring Multiple Myeloma and related diseases
Introduction
Did you know?
  • IMWG recommend serum free light chain (sFLC) testing to monitor myeloma patients when the monoclonal protein produced is not measurable by traditional techniques.
  • Evidence suggests the inclusion of sFLC testing in monitoring all patients, even those with measurable disease. This enables earlier detection of disease relapse, allowing appropriate treatment plans to be initiated.
Find out more about why you should routinely monitor all myeloma patients with Freelite® assays
View our summary of publications
Speaker
Featured in this video
Image of speaker, Dr. Aurore Lison
Dr. Aurore Lison
Medical Science Liaison, Binding Site, part of Thermo Fisher Scientific
Key Takeaways
44% of myeloma patients relapse with a different clone
Changes in monoclonal protein expression occur in almost half of the myeloma patients between diagnosis and relapse. Zamarin et al. studied the monoclonal protein type for 126 myeloma patients at both diagnosis and relapse using SPE and serum Freelite tests.
Overall, the study observed that the monoclonal protein type at relapse differed to the type at diagnosis in 44% of all patients, that 56% of the patients express FLCs at relapse and that 22% of the patients undergo FLC escape.
For the different myeloma types, Zamari et al. found that:
  • Among patients that expressed both FLC and intact immunoglobulins at diagnosis, 22% relapsed with light chain only (FLC escape).
  • Among patients that expressed only intact immunoglobulins at diagnosis, 17% patients started also to produce FLC at relapse.
sFLC testing allowed these light chain escapes and relapses to be detected. These may have been missed if testing was performed with SPE alone.
50% of light chain escape patients develop renal impairment
Light chain escape is linked to significant morbidity and mortality, with 50% of patients developing renal impairment. Brioli et al. investigated patient survival according to their monoclonal protein relapse type. The study observed that patients relapsing with FLC involvement experienced poorer prognosis and overall survival compared to intact immunoglobulin only patients.
Download our summary of publications to find out more about why you should routinely monitor all myeloma patients with Freelite assays.
If you liked this content, you will also like these:
Freelite® assays
Product
Freelite® assays
Detect and Monitor Multiple Myeloma with Freelite assays
Myeloma Monitoring Guidelines
Video
Myeloma Monitoring Guidelines
Listen to Dr Claudia Fogl to find out more about guidelines for monitoring Multiple Myeloma
References
  1. Zamarin D et al. Patterns of relapse and progression in multiple myeloma patients after auto-SCT: implications for patients' monitoring after transplantation. Bone Marrow Transplant. 2013 Mar;48(3):419-24.
  2. Brioli A et al. Serum free immunoglobulin light chain evaluation as a marker of impact from intraclonal heterogeneity on myeloma outcome. Blood. 2014 May 29;123(22):3414-9.
The products referred to on this page may differ from what is commercially available in your country. To find out what is available in your country, please contact your local Binding Site representative.
Freelite is a registered trademark of The Binding Site Group Limited (Birmingham, UK)
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you