Complement assays
Unlocking the potential of complement testing in immune disorders
Overview
What are complement assays?
The complement system is a crucial component of our innate immune system that supports the body's defense against injuries and infections. Its functions include activation of the inflammatory response, phagocytosis-mediated destruction of bacteria and viruses, pathogen agglutination, and cell lysis of susceptible organisms.
There are three distinct pathways through which the complement cascade can be activated, each triggered by different mechanisms. These pathways involve more than 50 different proteins.
Complement testing is recommended for the assessment of specific complement disorders.1
Understand more about immunological disorders
Primary immunodeficiency, secondary immunodeficiency and inborn error of immunity
Range of assays
Comprehensive assay portfolio 
  • Comprehensive and optimized assays for quantification and functional assessment of complement. These assays are included in clinical guidelines2
  • The functional CH50 assay is used to measure classical complement pathway activity and can be used to investigate complement deficiencies
  • The complement component assays (C3, C4, C1 Inactivator, C1q, C2, C5 and Factor B†)* aid in the determination of immune deficiencies. 
*Product availability is subject to country specific regulatory requirements. Contact your local representative for availability in your country. Not for use in the U.S. or China. 
The legal manufacturer is TRIMERO diagnostics SL.
Primary immunodeficiency (PID)
Find out more about the warning signs of primary immunodeficiencies, the different manifestations and the best diagnosis practice
Primary immunodeficiency screening 
Protocol
E. de Vriesin collaboration with European Society for Immunodeficiencies (ESID) members developed a protocol to screen for primary immunodeficiency
Protocol for screening for Primary Immunodeficiency Disorders
Precision diagnostics 
Complement assays optimized for use with the Optilite® Analyser
Human complement functional assay – CH50  Human complement protein assay - C1 inactivator
This functional assay aids in the characterization of classical complement pathway activity, and therefore aids in the investigation for complement deficiencies. This assay may be used where there is a clinical suspicion of immunological disorders. This assay quantifies the levels of C1i and can therefore aid in the characterization of complement deficiencies. The assays are also cleared as an aid in the diagnosis of hereditary angioedema. 
Human complement protein assay - C3 Human complement protein assay - C4
This assay quantifies the levels of C3 and can therefore aid in the characterization of complement deficiencies. This assay could be used if there is a suspicion of immunodeficiencies or as further testing dependent on initial tests. This assay quantifies the levels of C4 and can therefore aid in the characterization of complement deficiencies. Inherited or acquired deficiencies in C1 inhibitor cause decreased serum levels of C4, which are associated with angioedemas.
Human complement protein assay - C1q Human complement protein assay - C2
This assay quantifies the levels of C1q and can therefore aid in the characterization of low complement activity in immunological disorders such as complement deficiencies, SLE and glomerulonephritis. This assay quantifies the levels of C2 and can therefore aid in the characterisation of low complement activity in immunological disorders such as, complement deficiencies, SLE and glomerulonephritis.
Human complement protein assay - C5 Human complement protein assay - Factor B†
This assay quantifies the levels of Complement C5 levels and can therefore aid in the diagnosis of abnormal metabolism of the proteins of the Complement System, and the consequent increase in susceptibility to infections. This assay quantifies the levels of Factor B and can therefore aid in the diagnosis of abnormal metabolism of the proteins of the Complement System, and the consequent increase in susceptibility to infections.
Image of a diagram showing a liposome based CH50 Method
Optilite® CH50 enhanced productivity 
  • Automated – Quantifiable, quality results to avoid subjective interpretation
  • Optimized - Consistent, reliable results instills clinical trust
  • Fast – Turn around time of ~15 minutes to deliver timely clinical care
  • On board reagent stability of 30 days helps save on reagent cost
  • Shows good correlation when compared with conventional methods
  • Defined reference limits and measuring ranges for serum and plasma for user convenience 
Optilite is a registered trademark of The Binding Site Group Ltd (Birmingham, UK) in certain countries.
Photo of a doctor showing a patient a device screen
Liposome-based CH50 method 
  • Automated method for quick functional assessment of the classical complement pathway
  • Uses antigen-coated liposomes encapsulating glucose-6-phosphate dehydrogenase (G6PDH) enzyme which are further sensitized with antibodies
  • When reagents are mixed with the clinical sample, complement in the sample is activated by Ag/Ab complexes on the liposome surface, resulting in liposome lysis and the release of G6PDH
  • G6PDH then reduces NAD to NADH, causing a measurable change in absorbance proportional to the complement activity in the sample
Immune Disorders
Immune Disorders
Immune Disorders
Learn more about the immune disorders that our complement assays aim to help diagnose and monitor.
Optilite® Analyser
Instruments
Optilite® Analyser
Transform your diagnostics with Optilite® - a fully automated, robust benchtop analyser for special protein testing.
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you
References
  1. Lung, Thomas et al. “The utility of complement assays in clinical immunology: A comprehensive review.” Journal of autoimmunity vol. 95 (2018): 191-200.
  2. Antonis Fanouriakis et al. Ann Rheum Dis 2021;80:14-25