  • Contact
Accelerating ScienceBehind the Bench / Pharmacogenomics / Biobanks in Northern Europe – The Future of Predictive Genomics
Biobanks in Northern Europe – The Future of Predictive Genomics
Written by Behind The Bench Staff | Published: 11.16.2021
Biobanks in Northern Europe and the Future of Predictive Genomics
Complexity and Data Requirements of Biobanks
Disease Risks and Pharmacological Possibilities
Dataset must contain genomic data on enough people to achieve statistical relevance
Genetic and non-genetic medical conditions must be taken into account
Predictive genomics holds tremendous promise, but realizing that promise has been a fitful process. A dataset that can properly inform medical providers about a person’s disease risks and pharmacological possibilities has to contain genomic data on enough people to achieve statistical relevance and information about those people’s other medically relevant statistics, including those that are not genetically determined. This is the premise of a biobank, and the biobanks in Finland and Estonia are already revealing just how much can be learned when whole countries put in the resources necessary to use genomic information to its fullest potential.
Finland and Estonia – Examples of Successful Biobanks
Finland and Estonia are both highly developed countries with small, relatively homogenous populations and universal healthcare. Combined with their advanced health-records systems and robust understanding of informed consent, both countries were ideal testbeds for demonstrating the benefits of a thorough biobanking system. FinnGen, the Finnish biobank, and its Estonia counterpart collected both genomic and medical information from sizable fractions of each country’s population—roughly 10% of Finland and 20% of Estonia—and combined it with medical test results and other medical records available through their respective healthcare systems to create comprehensive databases of each participant’s health information.
Related: Massively Parallel Genetic Testing of the Han Chinese Population in Taiwan
Genome-Wide Association Studies
In Finland, this information is used as part of genome-wide association studies (GWAS) to identify gene variants of interest for various health conditions, including breast cancer, using Thermo Fisher Scientific GWA Arrays specifically designed for FinnGen’s research. This information, in turn, can be used to assign polygenic risk scores (PRSs) based on variants of concern present in a person’s genome and to act accordingly, such as performing direct medical tests more often on especially susceptible people.
Helping Doctors with Diagnosis & Choosing Treatments
In Estonia, this work is being used to help develop a tool to aid doctors with diagnosis and especially with choosing treatments for complex conditions like depression. Personalized genetic information may yet spare more Estonians from the difficult and sometimes life-threatening process of trial and error currently needed to find the right medication for conditions like these.
Webinar: Predictive Genomics Insights from Experts
In this webinar, Dr. Aarno Palotie of the Institute for Molecular Medicine Finland (FIMM) at the University of Helsinki and Dr. Andres Metspalu of the Estonian Genome Center ad the University of Tartu discuss their countries’ respective biobanks: what tools were used to collect genomic information, what other medical information they encompass, and how they have already been used to assist with medical practice.
Sign up to watch the webinar to see the full story or click here to learn more about predictive genomics.
Related: What Does it Take to Build a Predictive Genomics Program?
Share this article
55
SHARES
FacebookLinkedinTwitterMail
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Research Use Only (RUO) Recombinant Antibodies for Biotech and Pharmaceutical Research
What Does it Take to Build a Predictive Genomics Program?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook