  • Contact
Accelerating ScienceBehind the Bench / Forensics / Evidence Found of Jewish Holocaust Victims at the Nazi Death Camp in Sobibór
Evidence Found of Jewish Holocaust Victims at the Nazi Death Camp in Sobibór
Written by Behind The Bench Staff | Published: 12.15.2021
It is estimated that 11 million people, 6 million of Jewish descent, were killed during the Holocaust of World War II.1 In a recent survey,2 it was found that two-thirds of young American citizens  (18-39) were unaware of this tragedy and almost half could not name a single concentration camp.  Clearly, we need to do more to educate the world on this tragedy which occurred just two generations ago.
Diepenbroek et al. recently published a paper citing evidence of Jewish remains at the Sobibór death camp in Poland. Ten sets of remains were discovered that were originally assumed to be from Polish victims of the communist regime. However, DNA analysis revealed that they were, instead, from Ashkenazi Jews who were murdered by the Nazi regime.3
Sobibor camp map
Lineage marker analysis was performed on both haploid marker systems, mitochondrial and Y-chromosomal DNA. The two independent laboratories involved in this study (Szczecin, Poland, and Innsbruck, Austria) performed initial Sanger as well as confirmatory and extending mitogenome Massively Parallel Sequencing using the Applied Biosystems Precision ID mtDNA Whole Genome Panel.  The Y-chromosomal lineages were investigated using the Applied Biosystems YFiler Plus PCR Amplification Kit, their haplogroup status was estimated with the Nevgen Y-DNA Haplogroup Predictor and confirmed by direct Sanger sequencing of the signature mutations. The analysis of both maternal and paternal lineage markers indicated that these remains were similar, is some cases even indistinguishable, to lineages found within the Ashkenazi Jewish population. After this discovery, the remains were reburied, following prescribed Jewish rituals, at the place of their discovery.
Please read the open access article for more on the history and excavation of the  Sobibór camp, along with the details of the genetic analysis methods. And you can watch Dr. Diepenbroek in this webinar as she discusses her study.
You can learn more about integrated human identification solutions and services, and how these can assist in helping you achieve your goals.
For research, forensic or paternity use only. Not for use in diagnostic procedures.
Sources:
1. https://www.washingtonpost.com/opinions/11-million-not-6-million-died-in-the-holocaust/2017/05/26/6fdcc270-3f1c-11e7-b29f-f40ffced2ddb_story.html
2. https://www.theguardian.com/world/2020/sep/16/holocaust-us-adults-study
3. https://genomebiology.biomedcentral.com/articles/10.1186/s13059-021-02420-0
Share this article
67
SHARES
FacebookLinkedinTwitterMail
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Your Vital Research Data is Secure with Thermo Fisher Connect
Unsolved Cases: Lawfully Owed DNA May Hold the Answers
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook