  • Contact
Accelerating ScienceBehind the Bench / General / Raw material considerations for cell therapies
Raw material considerations for cell therapies
Written by Behind The Bench Staff | Published: 12.16.2022
Raw material selection for cell therapy manufacturing can have a significant impact on the safety and efficacy of the final product.  The input materials used in the manufacturing process can typically be classified into one of two categories:
  • Raw materials – Materials used in manufacture of but not intended to be present in the final product – e.g., cell-culture media, growth factors, supplements, cytokines, cell-separation reagents. Also known as “ancillary materials” by US regulations.1
  • Excipients – Materials administered as part of the product, help maintain quality attributes of the cells – e.g., electrolyte solutions, cryopreservatives.
As definitive regulations have not yet been developed for these materials, determining a qualification regime for the selection of raw materials (RMs) can be challenging and confusing for cell therapy manufacturers.
To clarify these issues, this blogpost intends to provide a high-level overview of the considerations used to select raw materials that mitigate risk and align with current regulatory guidelines.
For more information, check out the accompanying chapter on raw material considerations (chapter 1) in our Cell Therapy Handbook.
What Type of Qualification is Required for Ancillary Materials?
To date, no specific cGMP guidance exists for raw material manufacturing2—suppliers may say their products are manufactured under cGMP conditions, with claims ranging from declarations of cGMP based on following a subset of cGMP guidelines; independent quality management system certification (e.g., ISO9001); or even regulatory agency inspection if the site is manufacturing regulated products.
One of the most globally recognized raw material guidance documents, USP <1043>, provides a framework for classifying raw materials into four different tiers based on risk.
raw material risk categories
Figure 1. USP <1043> raw material risk categories
Summary
A cell therapy’s manufacturing components are critical to developing a reproducible and safe product. Proper sourcing of materials early in development of a cell therapy from reliable suppliers who make products specifically for cell therapeutics can shorten the development timeline, dramatically reduce costs, and improve the likelihood of approval from regulatory authorities.
Additional resources
This article provides an overview of the numerous challenges and considerations that a cell therapy manufacturer must address while selecting appropriate raw materials. For more in-depth discussions on these topics, we recommend reading up on the accompanying chapter in our Cell Therapy Handbook.
References
  1. Thermo Fisher Scientific (2020) Manufacturing pluripotent cell therapeutics
  2. USP (2006) General Chapter <1043>: Ancillary Materials for Cell- and Tissue-Based Products. In: USP-NF English Edition. Rockville: United States Pharmacopeial Convention.
Share this article
FacebookLinkedinTwitterMail
Your IVDR Transition, Now With the 3500-Series Genetic Analyzers
Cell isolation, engineering, and expansion
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook