  • Contact
Accelerating ScienceInside Biobanking / Case Studies / Next-Generation DNA Sequencing: Predictive Biomarkers for Cancer Treatment
Next-Generation DNA Sequencing: Predictive Biomarkers for Cancer Treatment
Written by Melissa J. Mayer | Published: 05.02.2017
DNA sequence / Abstract background of DNA sequence. Image: enzozo/Shutterstock.com.As treatment options for cancer patients increase, oncologists stress the importance of identifying biomarkers capable of predicting treatment outcomes. Unfortunately, biomarker discovery is complicated by the heterogeneous nature of tumors themselves. Overcoming this obstacle requires the acquisition of high-quality tumor samples collected prior to treatment, preferably with follow-up data demonstrating treatment outcomes. Ideally, these samples should be fresh-frozen for next-generation DNA sequencing (NGS) as well as proteomics and metabolomics techniques.
Bins et al. (2016) recently described a multicenter initiative designed to address the safety of image-guided tumor biopsy procedures followed by DNA sequencing for treatment-predictive biomarker discovery.1 To do this, the research team implemented an “umbrella” biopsy protocol, recruiting patients with all solid tumor types, poised to begin varied treatment protocols. These recruits (n = 450, the majority diagnosed with breast cancer, lung cancer, colorectal cancer or melanoma) underwent percutaneous biopsy guided by ultrasonography, computed tomography or endoscopy. The clinicians at three centers in the Netherlands collected two to four core biopsy specimens per subject, which they immediately snap-froze for shipment to the Center for Personalized Cancer Treatment (CPCT). There, researchers subjected the samples to histological analysis and DNA sequencing. Of the biopsied patients, 324 (72%) received systemic therapy after biopsy. For most, this treatment included targeted agents. Clinicians reported post-biopsy adverse events for 2.1% of the biopsies (n = 10). These included pain (n = 4), hypertension (n = 1), vasovagal reaction (n = 1), pneumothorax (n = 3) and pleural hemorrhage (n = 1). The CPCT reported that most (81%) of the biopsy samples had a tumor cell percentage of at least 30%, and 91% of these contained sufficient DNA (500 ng) for sequencing. Overall, 331 of the specimens (74% of image-guided biopsies) met preset criteria for DNA sequencing and biomarker discovery. The researchers provided sequencing data for 73 of these biopsy specimens and reported the following most frequently mutated genes across tumor types: TP53, APC and BRAF. They indicated that their sequencing data for these specimens is concordant with the Cancer Genome Atlas. They further disaggregated the mutated gene data by histological origin.
Table 1. Total mutated genes
Mutated gene
Total specimens (n = 73)
TP53
34 (47%)
APC
15 (21%)
BRAF
13 (18%)
CCNB3
12 (16%)
LRP2
12 (16%)
ATRX
11 (15%)
CTBP2
11 (15%)
FAT3
11 (15%)
KRAS
11 (15%)
OBSCN
11 (15%)
PIK3CA
10 (14%)
In summary, Bins et al. offer this study as evidence that biobanking image-guided biopsy samples derived from multiple collection centers is a feasible undertaking. Further, they indicate that these biopsies are generally well tolerated by patients with advanced cancer. They suggest that their framework (fresh-frozen samples from biopsies taken prior to systemic treatment, suitable for NGS as well as proteomic and metabolomic analyses) could help future endeavors to collect tumor tissue and correlate the arising molecular data with clinical outcomes. Ultimately, this undertaking could assist in the identification of reliable predictive biomarkers for cancer treatment.    
Reference
1. Bins, S., et al. (2016) “Implementation of a multicenter biobanking collaboration for next-generation sequencing-based biomarker discovery based on fresh frozen pretreatment tumor tissue biopsies,” The Oncologist [Epub ahead of print], doi:10.1634/theoncologist.2016-0085.
Share this article
FacebookLinkedinTwitterMail
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Keeping It Cool: Protecting Cryopreserved Samples from Degradation
The Effect of Preanalytical Variables on Plasma Samples Stored in Biobanks
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook