  • Contact
Accelerating ScienceExamining Food / Expert Insight / General Topics / Examining the Flavorings of Earl Grey Tea
Examining the Flavorings of Earl Grey Tea
Written by Emily Humphreys | Published: 04.29.2014
The European Union maintains a list of authorized flavoring substances as a result of evaluations by the European Food Safety Authority. Flavorings are regulated to ensure dietary exposure is maintained at a safe level. In a recent publication, researchers sought to demonstrate the distribution of flavorings found in food products. “Earl Grey” tea was chosen for this investigation 1
Bergamot oil is one of the key flavorings found in Earl Grey tea. A previous study by Dugo et al.2 characterized bergamot oil and fount that it contained linalyl acetate(15.6–41.4%), linalool (1.7–26.0%) and limonene (10.5–45.8%) as the major constituents; along with with β-pinene (0.08–11.0%) and γ-terpinene (3.7–11.4%). The distribution of flavorings found in bergamot oil can vary greatly based on the origin, latitude, growing year and harvest period.
The researchers purchased 90 tea samples in 10 European Union member states that included: Germany (n = 28), Hungary (n = 10), Poland (n = 10), United Kingdom (n = 10), Italy (n = 6), Spain (n = 6), Denmark (n = 5), Finland (n = 5), France (n = 5) and Ireland (n = 5). The purchased tea samples included samples from international/national brands (n = 52), private label brands (n = 16) and specialty tea shops (n = 22). This study also surveyed teas packaged in tea bags (n = 54) as well as loose leaved teas (n = 36).
The researchers used liquid-liquid extraction with hexane to separate the flavoring substances from the tea. Once separated, they dried and diluted the tea. They were able to determine the flavoring contents using capillary gas chromatography (GC) with a GC 8000 series gas chromatograph (Thermo Scientific) with on-column injection. Chromcard software (Thermo Scientific) was used to acquire the data.
The researchers determined linalyl acetate, linalool and limonene were consistently present as the major flavoring substances. They analyzed the enantoiomeric compositions of linalyl acetate, linalool, limonene, and β-pineneusing by multidimensional capillary GC analysis. This analysis revealed that the enantiomeric purities of the flavoring substances had no significant impact on the contents of the flavoring substances median concentrations of linalyl acetate (p = 0.07632) and linalool (p = 0.4385).
The amounts of linalyl acetate and linalool present in the teas remained fairly consistent from country to country, and no significant differences were found (linalyl acetate: p = 0.459; linalool: p = 0.347).
When the researchers looked for differences in flavorings based on how the teas were packaged, they found teas sold as loose leaves had a higher median content of linalyl acetate and linalool (66% and 39%, respectively), and this was higher than in teas packaged in tea bags. They also noted that the flavoring substances degraded over time, especially teas that were not flavored with granulates.
The researchers were able to conclude that the flavorings found in Early Grey teas contain a low level of uncertainty and that they fall within accepted levels based on dietary exposure assessments authorized by  the European Union.
References
1. Orth, A.M., Yu, L., & Engel KH. (2013) “Assessment of dietary exposure to flavouring substances via consumption of flavoured teas. Part 1: occurrence and contents of monoterpenes in Earl Grey teas marketed in the European Union.”, Food Additives and  Contaminants Part A Chemistry Analysis Control Exposure Risk Assessment.;30(10) (pp. 1701-14). doi: 10.1080/19440049.2013.817687 26.
2. Dugo G, et al. (2012) “Characterization of cold-pressed and processed bergamot oils by using GC-FID, GC-MS, GC-C-IRMS, enantio-GC, MDGC, HPLC and HPLC-MS-IT-TOF.” Journal of Essential Oil Research, 24 (pp. 93–117)
Share this article
5
SHARES
FacebookLinkedin
package nutritional facts
Content and Label Must Match – Especially When It’s On the Front of the Package
This year, the US Food & Drug Administration (FDA) issue... by Kimberly Durkot / 05.13.2025
Read More
packaged rice cakes
Does Orientation Matter to a Food Metal Detector?
Editor’s Note*:  We received an inquiry from a a read... by Kimberly Durkot / 05.06.2025
Read More
A granola bar in a wrapper
A Good Reason for Slacking when it Relates to Checkweighers
Checkweighers are essential tools used in various industries... by David Prieto / 01.28.2025
Read More
Cranberry Sauce / Jellied Cranberries
The Importance of Portion Control on Thanksgiving — For Food Manufacturers
It’s that time of year when holidays are approaching a... by Kimberly Durkot / 11.26.2024
Read More
Tips for Successful Culture Media Preparation
Download Now
Guide to Metal Detection and X-ray Inspection of Food
Download Now
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Fact sheet on Staphylococcus
Surveying the Fluoride Content in Soft Drinks and Other Beverages
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook