  • Contact
Accelerating ScienceExamining Food / Packaged Product Inspection / Infographic: Is X-ray Inspection of Packaged Food Safe?
Infographic: Is X-ray Inspection of Packaged Food Safe?
Written by Bob Ries | Published: 09.01.2015
Xray FoodsFinding physical contaminants in food products is critical to the general health of the public. X-ray inspection is one of the most effective ways to keep products safe.  But some people are under the false impression that it may be a health hazard.
Food X-ray inspection systems do not use radioactive materials to generate X-rays. Instead they use X-ray tubes that are run at very high voltage where electrons are accelerated across a gap bombarding a tungsten material to generate images. When the tube is turned off no X-ray energy is emitted.
On average, Americans receive a radiation dose of about 0.62 rem [620 millirem (mrem)] each year. Half of it comes from natural background radiation.* International Standards allow exposure to as much as 5,000 mrems a year for those who work with and around radioactive material. According to the U.S. Food and Drug Administration (FDA) website: There are no known adverse effects from eating food, drinking beverages, using medicine, or applying cosmetics that have been irradiated by a cabinet X-ray system used for security screening. 
Take a look at this infographic and get a quick glimpse at the average doses of radiation people receive from exposure to common products/situations.
*Resources: US Nuclear Regulatory Commission: http://www.nrc.gov/about-nrc/radiation/around-us/doses-daily-lives.html and http://www.ans.org/pi/resources/dosechart/.
Share this article
7
SHARES
FacebookLinkedin
package nutritional facts
Content and Label Must Match – Especially When It’s On the Front of the Package
This year, the US Food & Drug Administration (FDA) issue... by Kimberly Durkot / 05.13.2025
Read More
packaged rice cakes
Does Orientation Matter to a Food Metal Detector?
Editor’s Note*:  We received an inquiry from a a read... by Kimberly Durkot / 05.06.2025
Read More
A granola bar in a wrapper
A Good Reason for Slacking when it Relates to Checkweighers
Checkweighers are essential tools used in various industries... by David Prieto / 01.28.2025
Read More
Cranberry Sauce / Jellied Cranberries
The Importance of Portion Control on Thanksgiving — For Food Manufacturers
It’s that time of year when holidays are approaching a... by Kimberly Durkot / 11.26.2024
Read More
Tips for Successful Culture Media Preparation
Download Now
Guide to Metal Detection and X-ray Inspection of Food
Download Now
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
What Rot! Strawberry Proteomics and the Art of Staying Fresh
Food Safety Predictive Modeling
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook