  • Contact
Accelerating ScienceExamining Food / Packaged Product Inspection / Inspecting Snack Foods Through Metalized Film Packaging
Inspecting Snack Foods Through Metalized Film Packaging
Written by Bob Ries | Published: 05.26.2015
potato chip metalized bagCommon package types for snack foods including chips, crackers, bars, pretzels, nuts and others is a metalized material that is automatically formed into a bag and has heat seals on either end. Folding cartons, composite cans, flexible material laminations and other packaging alternatives are also found in the snack foods category. 
Although most snack foods don’t have the product effect issues found in baked items (like warm, moist, and dough stages), spotting contaminants in metalized film that uses a vertical form-fill-seal (VFFS) machine or flow wrapper is a challenge. According to the Institute of Food Technologists, Metallized films are plastics containing a thin layer of aluminum metal…. These films have improved barrier properties to moisture, oils, air, and odors, and the highly reflective surface of the aluminum is attractive to consumers.
While these types of food packaging products are great for keeping out contaminants, they’re not good candidates for inspection using metal detectors. Metal detectors use high frequency radio signals (50 – 700 KHz) to detect the presence of metal in food or other products. A digital signal processing system analyzes certain signals and sends an alert if metal is present and could cause a food hazard or safety issue.
Even with drop-through metal detection technology, there typically is not enough room in the VFFS machine to mount a reject mechanism. Therefore, there is no easy way to reject a single product. (A possible work around is to give a signal to the bagger to make a double bag when a contaminant is found. The double bag, coupled with an audio alarm, signals the person packing the case that there has been a metal hit.)
With these process and material obstacles in place, X-ray equipment is the better solution to address snack food inspection challenges. X-ray systems produce density images that are analyzed for irregularities. X-ray inspection is one of the first lines of defense to identify the presence of foreign contaminants in food products before they have the chance to leave the processing plant. Unlike metal detectors that offer protection from many types of metal contaminants encountered in food production, X-ray systems can ‘ignore’ the packaging and find virtually any substance that is denser or sharper than the object containing it.
Additionally, in the case of bars and other similar snack products, X-ray inspection can be used for spotting missing or broken pieces.
For more information, read the Market Insights And Solutions About Packaged Baked Goods & Snack Products white paper and visit our Food Community site.
Share this article
4
SHARES
FacebookLinkedin
packaged rice cakes
Does Orientation Matter to a Food Metal Detector?
Editor’s Note*:  We received an inquiry from a a read... by Kimberly Durkot / 05.06.2025
Read More
Cranberry Sauce / Jellied Cranberries
The Importance of Portion Control on Thanksgiving — For Food Manufacturers
It’s that time of year when holidays are approaching a... by Kimberly Durkot / 11.26.2024
Read More
Tips for Successful Culture Media Preparation
Download Now
Guide to Metal Detection and X-ray Inspection of Food
Download Now
Comments
  1. bblfoods says
    December 24, 2019 at 10:35 am
    We at BBL Foods, support and supply of hardly used batch type potato chips machines with an excellent working condition based on the capacity requirements.
    Reply
  2. Bblfoods says
    May 30, 2021 at 5:12 am
    Thanks for this post….at BBL Foods, support and supply of hardly used batch type potato chips machines with an excellent working condition based on the capacity requirements.
    Reply
  3. Bblfoods says
    August 1, 2021 at 4:45 am
    Thanks for this post.. We at BBL Foods, support and supply of hardly used batch type potato chips machines with an excellent working condition based on the capacity requirements.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
No Cavities Found With These X-Rays
Biofilms: Mechanisms and Control Strategies for Foodborne Pathogens
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook