  • Contact
Accelerating ScienceExamining Food / Chemical and Elemental Analysis / Surveying the Fluoride Content in Soft Drinks and Other Beverages
Surveying the Fluoride Content in Soft Drinks and Other Beverages
Written by Emily Humphreys | Published: 05.01.2014
Fluoride is naturally occurring in low levels in water, soil, plants, and animals. In recent years, studies have reported on the health benefits (especially in children) of supplementing with fluoride. Ingesting fluoride strengthens tooth enamel and prevents tooth decay.1
However, children are especially susceptible to high amounts of fluoride, and exceeding safe amounts (3-4mg/day for adults, and 1.5mg for children) can leading to dental and skeletal fluorosis and even damage to major organs. There is some concern that the amounts of fluoride consumed may reach dangerous levels when fluoridated water is used to manufacture soft drinks, teas, juices and other beverages. A research group from Portugal has recently investigated the fluoride content of these beverages by performing a potentiometric analysis.2
The researchers obtained 183 samples of soft drinks, juices, nectars, juice drinks, concentrates, teas and infusions marketed in Portugal. They included a large number of products and brand names, and they paid particular attention to products marketed for children or infants.
This investigation was performed using a fluoride combination ion-selective electrode (Thermo Scientific) and a potentiometer selected for fluoride analysis. After carefully optimizing and validating the potentiometric method, the researchers were able to determine the fluoride content in the target samples.
Portugal does not have legislation or requirements for the experimental recovery of fluoride; however, Portugal does have a national legislation requirement for a parametric value of 1.5 mg/L and a maximum RSD (repeatability of the standard deviation) of 10% and minimum recovery of 20%. By spiking control solutions with known quantities of fluoride, the researchers determined their methods had an overall RSD ≤ 10%.
When these methods were applied to the samples of beverages, the researchers were able to determine all samples had fluoride greater than the minimum level of quantification of (0.06 mg/L). The greatest amount of fluoride was found in soft drinks and one extract-based soft drink sample contained 2.0 mg/L of fluoride. Most extract-based samples contained less fluoride, with levels as low as 0.10mg/L. The researchers measured the mean amount of fluoride in carbonated soft drinks at 0.18 mg/L). The juice-based soft drinks had fluoride levels between 0.10 and 1.3 mg/L with a mean of 0.25 mg/L.
Fruit juices had a low fluoride concentration at 0.01– 0.29 mg/L (0.37 ± 0.11 mg/L) and ice teas had a higher fluoride concentration (1.28 mg/L) which is consistent with a previous study3. The smallest amount of fluoride was found in infusions (0.12 ± 0.01 mg/L) and teas (0.16 ± 0.12 mg/L).
Because some of these beverages (especially the extract-based soft drinks) contain considerable amounts of fluoride, the researchers suggest the importance of being aware of the amount of fluoride exposure. They also recommend that further experiments focus on determining the fluoride exposure in the Portuguese population as a whole.
References:
1. Browne D, Whelton H, O’Mullane D. (2005) “Fluoride metabolism and fluorosis.”, Journal of Dentistry. Mar;33(3):177-86. Epub 2004 Dec 9.
2. Fojo C, Figueira ME, & Almeida CM (2013) “Fluoride content of soft drinks, nectars, juices, juice drinks, concentrates, teas and infusions marketed in Portugal.”, Food Additives and Contaminants Part A, 30(4):705-12. doi: 10.1080/19440049.2013.785636.
3. Opydo-Szymaczek J, Opydo J. (2010) “Fluoride Content of Beverages Intended for Infants and Young Children in Poland.”, Food and Chemical Toxicology, Oct;48(10):2702-6. doi: 10.1016/j.fct.2010.06.043
Share this article
206
SHARES
FacebookLinkedin
package nutritional facts
Content and Label Must Match – Especially When It’s On the Front of the Package
This year, the US Food & Drug Administration (FDA) issue... by Kimberly Durkot / 05.13.2025
Read More
packaged rice cakes
Does Orientation Matter to a Food Metal Detector?
Editor’s Note*:  We received an inquiry from a a read... by Kimberly Durkot / 05.06.2025
Read More
Cranberry Sauce / Jellied Cranberries
The Importance of Portion Control on Thanksgiving — For Food Manufacturers
It’s that time of year when holidays are approaching a... by Kimberly Durkot / 11.26.2024
Read More
Tips for Successful Culture Media Preparation
Download Now
Guide to Metal Detection and X-ray Inspection of Food
Download Now
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Examining the Flavorings of Earl Grey Tea
Infrared Spectroscopy for Coffee and Banana Analysis – Renewable Biomass Fuels of the Future?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook