Thermo Fisher Scientific
Spectroscopy, spectrometry, and materials science stories and solutions about advancing research and improving product development
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAdvancing Materials / Breakthroughs in Materials Science / Dual-X: Breaking Down the Barriers to Chemical Analysis
Dual-X: Breaking Down the Barriers to Chemical Analysis
Written by Yuri Rikers | Published: 07.03.2019
Today, many researchers need fast information about the chemistry of their samples. Moreover, many work with materials sensitive to electron beams, making it difficult to obtain the data they need. The good news is that our new Dual-X detector solves both of these challenges.
To help researchers obtain faster 2D and 3D chemical information at lower doses on a wider range of samples, including beam-sensitive materials, we recently added two symmetrically positioned 100mm2 Racetrack detectors (Dual-X) to our Talos F200i scanning transmission electron microscope (S/TEM). This new feature addresses the difficulty of retrieving good quantitative data from non-symmetrical EDS arrangements and will allow scientists to characterize materials down to the (sub-)nanometer level in the fastest way.
The Dual-X detector was developed to deliver the highest analytical throughput, making it possible to perform energy-dispersive X-ray spectroscopy (EDS) analyses much faster than with previous EDS detectors. Large EDS maps can be generated in less than a minute.
Our high-speed Dual-X EDS detectors can be combined with the high-brightness X-FEG electron gun to maximize imaging and spectroscopy performance.
Dual-X detectors were used to produce this large-area, high-resolution EDS mapping of gold-nickel nanoparticles in less than one minute. (Sample courtesy J. Bursik, Institute of Physics of Materials, Brno)
Here are some of the benefits:
    • Symmetrically positioned Dual-X enables the highest analytical throughput and chemical analysis at lower doses on a wider range of samples.
    • Dual-X is embedded in our Velox Software, allowing users to easily acquire and analyze multimodal data for the most complete material characterization all in one tool.
    • Unique EDS absorption correction in Velox Software enables the most accurate quantification. Regardless of sample orientation, users always have access to the most accurate elemental information.
    • Dual-X is fully embedded and optimized for:
      • Fast, automated, unattended EDS tomography for 3D EDS enables instant access to your data
      • STEM and EDS Maps software for automated acquisition of statistically relevant data on large area images at high resolution. Data from different imaging and analysis modalities can easily be correlated including on-the-fly processing and statistics via Thermo Scientific Avizo software.
One of several EDS detectors configurable with the Talos, the Dual-X will be available starting this week. This new detector promises to help scientists answer questions that were previously unresolvable, taking automated 2D and 3D EDS to a new level.
To learn more about the Dual-X detector, please see our webpage or watch our video:
Yuri Rikers is Talos product marketing manager at Thermo Fisher Scientific.
To learn more about Dual-X detector, fill out this form to speak with an expert.
Share this article
FacebookLinkedinTwitterMail
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
A New Generation of Multiple Ion Plasma FIB Technology
Where Would We Be Without Our QA/QC Super Heroes?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook