  • Contact
Accelerating ScienceAnalyzing Metals / Precious Metals / 7 Questions about Gold Plating Answered
7 Questions about Gold Plating Answered
Written by Peter Anzalone | Published: 09.01.2020
7 Questions about Gold Plating AnsweredEditor’s Note:  After publishing an article by gold-plating expert and occasional guest writer, Calla Gold, we received several inquiries.  Calla was kind enough to answer the questions below.
The original article: How Gold Plating is Done, Step by Step
1. QUESTION 
“I have a very collectible and valuable vintage bracelet made of base metal with glass inserts. It has been gold plated once but the coat has worn off completely. If I am to gold plate the bracelet would it have to be taken apart first or can it be plated assembled?”
ANSWER 
Gold plating is an immersive process. It’s performed in a heated liquid bath. Your jeweler or plater would need to see the piece in person to determine if the glass needs to be removed. It’s not because of the glass itself, which is nonmagnetic and would be unharmed and left unplated. The potential problem is the glue, that may or may not have been used to create the piece. The plating solution might be contaminated by the glue. For me, that is what I worry about.
2. QUESTION 
I am a jewelry manufacturer. We face many difficulties in gold plating and rose gold plating in that it tarnishes very quickly. My clients complain that even 3 micron pieces tarnish in 2-3 weeks. For example, some of the rose gold plating goes black. My clients compare with similar jewelry they have purchased elsewhere that hasn’t tarnished. Can you help explain why there is an issue.
ANSWER
Different countries do different kinds of plating. Some platers claim they do micron plating, when many times (not all times) they do a “flash” plating. We have a customer that imports kilos and kilos of silver beads and asks us to redo the factory plating on a lot of them because of the thin plating and tarnishing.  
The three main issues I have seen with color plating are:
    1. Yellow gold or rose gold plated directly over silver. There should be a barrier of copper or palladium or nickel between the silver and the gold.
    2. Too thin a layer of plating.
    3. Too much copper in the rose gold plating mix, to make it pink. Copper tarnishes quickly and there’s a fine line in the chemistry of the plating baths. Too much copper and the rose gold will darken too quickly.
Some manufacturers are great at color plating and very thin plating. Others are great at clear coating and E coating. This makes their plating last almost forever, and it won’t change color, because the clear coating seals the piece of jewelry from the air. 
Hope that helps.
3. QUESTION 
How can you determine if a piece is real gold or other precious metal? Or if it’s plated or there’s lead in the jewelry?
ANSWER
XRF (X-ray fluorescence) technology is a non-destructive analytical technique used to determine the elemental composition of materials. There are portable XRF analyzers that can perform fast, accurate, and most importantly, nondestructive analysis of the precious metals. These gold spectrometers quickly provide the exact karat weight and percentages of all elements within an item – easily identifying non-standard, under-karated, and even advanced counterfeit gold with fire assay-comparable accuracy.  They can quickly distinguish between gold plating and solid gold, as well as determine the presence and concentration of other trace alloying elements and dangerous heavy elements (like lead) which could impact valuation and future refining needs. 
4. QUESTION 
Can the electroplating process be reversed?  I have circuit boards with gold on them.  Can I use this process to take the gold off the circuit board and deposit it on a piece of gold wire?
ANSWER
It is possible to use reverse current to plate off the gold from the circuit boards. However, other materials on the circuit boards may be plated off as well so if you want only pure gold plated on the wire, that may be a problem. If you are going to melt, refine or assay the wire then it shouldn’t be a problem.
5. QUESTION 
What happens to the plated gold after the old jewelry is cashed in?
ANSWER
A great deal of jewelry is made with just a layer of gold on the surface over another type of metal underneath. For example, gold vermeil jewelry uses sterling silver which has been gold plated. Some  gold-plated jewelry uses a base metal, such as copper or nickel, which is then electroplated with gold. For gold-plated items, their plating can vary in thickness, and no minimum value needs to be met for a piece of jewelry to be sold as gold plated.
The problem with trying to cash in these jewelry pieces is that the other alloying elements are not as valuable as gold, so the refiners need to separate them out and then evaluate just how much that heap of gold, other precious metals, and alloy materials is actually worth. (Hopefully the store operator who analyzed the jewelry pieces used an XRF analyzer to ensure the right value was assigned to the piece before giving out any cash to the customer.) Read more about assaying and assessing the jewelry. 
6. QUESTION 
I have a pair of earrings and I would like to have them gold plated. I have noticed a slight difference in color where the prong is soldered to the base of the earring. Will the gold plating cover the color difference making just that spot more uniform?
ANSWER
My short answer to the question is that gold plating is immersive and will be the same color regardless of subtle color differences on the underlying metal. With silver that has been gold plated, oxidation can cause the gold plating to darken in spots. If the other colored metal in your earrings is a metal that oxidizes, then it is possible that over time that oxidation could show up as darker, like a shadow in the gold plating.
7. QUESTION 
What’s the Difference Between Electroplating and PVD Coating?
ANSWER
See separate article on Calla Gold’s website: Gold Electroplating vs PVD Coating
Thank you to Calla Gold, a Santa Barbara Jeweler specializing in custom jewelry design and jewelry repair, and the author of Design Your Dream Wedding Rings From Engagement to Eternity, for answering our readers’ questions.
In addition, I suggest watching this excellent video on how plating works https://www.youtube.com/watch?v=PZquHLFVXvY 
Share this article
435
SHARES
FacebookLinkedinTwitterMail
gold metallic number 5
5 Precious Metals Analysis Methods
We’ve previously written about the four primary precious m... by Juan Pablo Zuluaga / 05.15.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. mahendra bamalwa says
    January 16, 2021 at 1:29 pm
    rosegold plating process
    Reply
  2. Eleni says
    April 6, 2021 at 7:24 am
    Gold plating simply does not work for me. While it’s not expensive, it stratches rather quick and you will get a worse looking jewel. I’d recommend buying a genuine golden alloy jewel or simply not plate it at all.
    Reply
  3. Dave Green says
    May 28, 2021 at 11:53 am
    That’s insightful!
    Reply
  4. Katherine Fontaine says
    September 10, 2021 at 3:09 am
    I’ve purchased gold plated and vermael in 14k and 24k. They fade, or my pH causes them (the underlying metal) to stink like dirty pennies. I’m learning and have done some “redipping” at home. So I have replated 2 gold plated cuffs for the “stink”. Both continue to smell once I’ve worn for a few minutes.. I know my pre process and adhered to them. I also plated in 14k for more than 15 minutes each. They look good, but …
    I’d buy solid or sterling but both pieces are engraved with meaning. Anyone else experience this? Solve it?
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
The 4 Methods for Analyzing Carbon in Steel: Which is Best?
Not All that Glitters is Gold
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook