  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / Brush Up on Your Stainless Steels
Brush Up on Your Stainless Steels
Written by Marlene Gasdia-Cochrane | Published: 12.12.2017
Brush Up on Your Stainless SteelsThere are more than 100 different grades of stainless steel, used in everything from surgical equipment to washing machines, to turbine blades. “Stainless” steel is actually a generic term referring to a variety of steel types, including Austenitic, Ferritic, and Martensitic. Stainless steel is made by adding varying amounts of chromium (Cr) and other alloying elements such as nickel (Ni) to iron and carbon to create a corrosion-resistant product. The types of alloys used depend on the final use of the product.  Does it need to be high and low temperature resistant? Flexible? Easily fabricated? Strong and durable? Easily cleaned and maintained?  Long lasting, with a low lifecycle cost? Aesthetically attractive? Environmentally friendly and recyclable?
The strength and hardness of stainless steel is an advantage in many applications, but the downside of adding strengthening alloys is poor machinability, meaning the material is difficult to cut and wears down the tooling. Processing the material through a cold rolling mill helps reduce thickness, increase strength and improve the surface finish. Hot rolling of steel occurs at very high temperatures, above the recrystallization temperature of the material, producing malleable sheets that can be formed in a variety of shapes. The process produces sheets that can be extremely long, which are then rolled into coils.
X-ray based metal coating weight and thickness sensors are used for accurate, real-time measurements to ensure the stainless steel is on spec, whether the steel is being hot or cold rolled.
Want to learn more about stainless steels?  We have compiled a list of our most-read articles about stainless steel and listed them below so you can access them all from one location.
What Is Stainless Steel? Part I
Read this introduction to the world’s most popular metal. Learn what it is made of, how it is used, and what the word ‘stainless’ really means when referring to a variety of steel types.
What is Stainless Steel? Part II: The Stainless Steel Extended Family
The multitude of stainless steel grades are grouped into five primary classifications. Find out which is the most widely used type of stainless steel, which has better corrosion, heat, and cracking resistance, and which is very hard and strong, though not as resistant to corrosion.
What Do Stainless Steel and Garlic Have in Common?
Tellurium is a metalloid, meaning it possesses the properties of both metals and nonmetals. It has a silvery-white appearance, and exhibits a metallic luster when pure. It’s an additive to steel, but can lead to a garlic odor on the breath upon human exposure.
PMI Testing of 347 Stainless Steel: As-Delivered vs. As-WeldedIn a competitive industrial climate, manufacturers must be vigilant about maintaining the quality of their products.  Even the smallest deviation from specifications of items like raw materials, fasteners, and welds can affect the quality of the product and the reputation of the company.
Why Surface Analysis Is the Key to Optimum Stainless Steel Performance
Ensuring a surface is free from contaminants is extremely important in guaranteeing that materials such as steel meet their desired performance specifications. Learn about an experiment and results using an XPS instrument to acquire elemental maps to identify the spatial distribution and chemical nature of the contamination on a stainless steel sample.
And if you want to take a look at a piece of stainless steel, just walk into your kitchen.  Most cutlery, knife blades, kitchen accessories, and cookware are made of this strong, corrosion resistant metal.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Lead Found in Pots Made from Recycled Scrap Metal
Thinking About the New iPhone X? Don’t Forget to Recycle Your Old Cell Phone. Here’s How…
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook