  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Buyer (and Seller) Beware…. Fake Gold Hitting Pawn Shops and Jewelry Stores
Buyer (and Seller) Beware…. Fake Gold Hitting Pawn Shops and Jewelry Stores
Written by Jonathan Margalit, PhD | Published: 12.05.2013
[Administrative Note: Because this is usually the busiest season for jewelers and pawnbrokers, this month we will be offering several articles that would be helpful to those who deal in precious metals. Today’s article addresses fake gold hitting pawn shops and jewelry stores.]
The livelihood of businesses that buy and recycle gold, such as jewelry stores and pawn shops, depends in part on being able to identify fake gold and to determine the accurate karat (K) weight of gold jewelry. With the volatility and high price of gold, failing to detect just a small variation in composition can be an expensive mistake. Even the seemingly simple task of distinguishing between gold plating and solid gold can be a challenge as counterfeiters become more adept.
Gold plated means that the jewelry is made with a layer of gold on the surface over another type of metal underneath. The most common identifying marks on gold plate are HGE (gold electroplate) and RGP (rolled gold plate), but not all gold-plated jewelry has such a stamp. High-quality gold plate can be thick enough to pass nitric acid scratch test, which only tests the surface layer. A deeper cut to the piece may yield more information about its content but will also damage it.
Other verification methods include weighing the jewelry in question against a known gold piece, or submerging the piece in water and measuring the displacement. Gold jewelry can be examined with a loupe to check for different colors under worn areas. Many jewelers also test with a strong magnet, as real gold isn’t magnetic.
Jewelers and pawn brokers that fail to verify the gold that comes into their shops using at least a couple of methods will fall prey to increasingly sophisticated counterfeiters who know how to get around these tests. Recent news stories describe scams involving jewelry made with non-magnetic metals that mimic gold, fakes plated with enough gold so as to appear to be all gold, or gold-plated or copper counterfeits stamped 14K or 18K.
“The fakes are getting better,” says one pawn shop owner. “They’re beating the magnet test, the weight test. Nowadays, gold needs to be tested several ways, including scratch testing or cutting into the metal, even if it means ruining the piece. Unfortunately, most of my customers intend to reclaim their jewelry and don’t want it damaged.”
To be absolutely certain of the composition of gold and other precious metals, many businesses are now turning to X-ray fluorescence (XRF) technology.  XRF analyzers provide a fast, accurate, and most importantly, a nondestructive method to test the purity and composition of all precious metals. XRF quickly provides the exact karat weight and percentages of all elements within an item – easily identifying non-standard, under-karated, and even advanced counterfeit material that acid testing is incapable of differentiating.  What’s more, some XRF instruments feature technology that can identify gold-plated items that, undetected, could adversely affect the profitability of your business. This functionality not only protects your interests but those of your clients, who can see for themselves the exact percentage of pure gold in their jewelry.
XRF analysis also can work in tandem with traditional testing to identify more heavily gold-plated items. When readings taken with an XRF analyzer on four or five different areas on a piece show differing results, this can be an indication that the item is heavily plated and further testing is warranted.  This would mean filing or drilling into the item to expose material beneath the surface layer.
See how XRF analysis compares with traditional acid testing in this 2-1/2 minute video.
Share this article
211
SHARES
FacebookLinkedinTwitterMail
radiation symbol for xrf safety
I Use XRF Analyzers on Metals… Is it Safe?
Do you use portable x-ray fluorescence (XRF) analyzers for... by Esa Nummi / 06.17.2025
Read More
gold metallic number 5
5 Precious Metals Analysis Methods
We’ve previously written about the four primary precious m... by Juan Pablo Zuluaga / 05.15.2025
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Maintaining Niton XRF analyzer
Enhancing Your XRF Instrument Investment: The Value of Preventative Maintenance
X-Ray fluorescence (XRF) analysis provides crucial elemental... by Tina Ferreira, Service Marketing Manager, and Sue Woods, Sr Manager of Product Development / 06.11.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Vincent says
    October 18, 2020 at 5:37 am
    I bought a piece of gold from a place called the piercing pagoda it’s in the tacoma mall and it says on the receipt 10k gold rope not plated gold rope and it’s plated now I payed almost 300$ for this piece of metal and they ripped me off. Now how I found out was I went to get a loan on the necklace only to find out it’s fake can I take legal action against them for this responses please thank u
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      October 19, 2020 at 12:50 pm
      Sorry, Vincent. We cannot comment on your experience, but it’s a good warning for others.
      Reply
  2. Jeffrey Olguin says
    June 15, 2021 at 6:13 am
    Yes you can take legal action against them for false advertising and a few other things. Such as defrauding the general public knowingly that the item(s) they are selling is not light. Keep your receipt as evidence your going to need it to prove your case. Also, if possible, get it in writing that your piece is fake from a jewelry or pawn shop. Good luck.
    Reply
Trackbacks
  1. Our Top 10 Analyzing Metals Articles - Analyzing Metals says:
    June 28, 2016 at 12:45 pm
    […] Buyer (and Seller) Beware…. Fake Gold Hitting Pawn Shops and Jewelry Stores The fakes are getting better. Jewelers and pawn brokers that fail to verify the gold that comes into their shops using at least a couple of methods will fall prey to increasingly sophisticated counterfeiters who know how to get around these tests. Recent news stories describe scams involving jewelry made with non-magnetic metals that mimic gold, fakes plated with enough gold so as to appear to be all gold, or gold-plated or copper counterfeits stamped 14K or 18K. Read here to find out how to be absolutely certain of the composition of gold and other precious metals..  […]
    Reply
  2. How Gold Plating is Done, Step by Step says:
    October 3, 2016 at 7:36 pm
    […] type of metal underneath. On some occasions, items are gold plated to try and fool buyers or cash-for-gold operators.  But for the most part, plating is done to enhance the look or wearability of a piece of […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Lead-Free by 2014: A Comparison of Tools for Compliance with the New Lead-Free Brass Mandate
XRF Technology Emerges as a Valuable Tool for Assessing Coins
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook