  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / Can XRF Analyze Metal Coating Thickness?
Can XRF Analyze Metal Coating Thickness?
Written by Esa Nummi | Published: 03.12.2015
transportation metal coatings analysisMetal is present in just about everything we use, from the cars and trains that get us to work to the computers and cell phones we use all day. In this blog, we talk about the myriad applications of metal, but an important thing that makes metal so incredibly useful is its coating.
Coatings impart valuable properties to metal: corrosion resistance, wear-resistance, solderability, machinability, hardness, brightness, adhesive qualities, and lubricity (reduced friction in moving parts). Industries that rely on coatings include automotive, aerospace, medical device, construction, and electronics. Metal is coated with a variety of substances, including chrome, nickel, and zinc.
Chrome: Chromeworks well with many substrates. Decorative chrome is used to improve appearance, while hard, or industrial, chrome offers excellent corrosion-, wear- and friction-resistance. Hard chrome is used in the nuclear, aerospace, and automotive industries as well as in the manufacture of many tools and medical devices.
Nickel: Nickel is a multi-purpose coating material; it’s aesthetically pleasing, corrosion-resistant, wear-resistant, stress-resistant, and magnetic. According to an article on Product Finishing, nickel can be made to plate with little or no stress so it’s especially useful for electroforming or aerospace applications. Nickel is used for engineering applications to promote wearability, and it provides a barrier layer on coins, jewelry and circuit boards. On strip steel and in aerospace applications, nickel is used for low stress or for resizing.
Zinc: Zinc works with a variety of substrates but is most often used to coat steel for protection against corrosion. There are many ways to apply zinc coating to steel but the most common are hot-dip galvanizing and continuous galvanizing.
The most common coating process is conventional electroplating. Other processes include electroless plating, galvanized coating (typically applied to steel, iron, and aluminum), physical and chemical vapor disposition, anodizing, mechanical coating, pickling, etching, and sputtering.
Why Is Coating Thickness Measurement Important?
Coating materials must be applied in certain thickness specifications in order to express the desired characteristics. Metals that are either over- or under-plated won’t perform as expected. Over-plating also wastes expensive coating material. If coating thickness inaccuracies aren’t detected, entire coils may end up being scrapped. Read Are You Wasting Expensive Steel Coating Material? to learn more.
Handheld XRF: Welcome to the Coatings Thickness Measurement Market
Coating thickness measurement instruments are available as on-line process measurement systems for metal manufacturing. Handheld, application-specific tools are commonly used on finished goods. A lesser-known, but extremely efficient technology for spot analysis of metal coatings in any application is portable X-ray fluorescence (XRF). XRF is a non-destructive analytical technique that measures the fluorescent (or secondary) x-ray emitted from a sample when it is excited by a primary x-ray source. The strength of the signal can be used to determine the thickness of the coating; a second reading can determine the thickness of the substrate. Handheld XRF instruments can be used anywhere in the supply chain, from incoming inspection to confirmatory testing of completed subassembly parts, to final product quality control. XRF coating thickness measurement method is covered by international standards ASTM B568-98  and ISO3497:2000.
Our next two articles will address metal coatings in the aerospace and automotive industries.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
16
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Arnoldo says
    September 22, 2016 at 8:38 pm
    Hi mates, pleasant post and pleasant urging commented
    here, I am truly enjoying by these.
    Reply
  2. Bellacelle review says
    September 23, 2016 at 4:18 am
    I know this site provides quality dependent articles
    and extra information, is there any other site which gives these kinds of stuff in quality?
    Reply
  3. Loyd says
    September 27, 2016 at 3:32 pm
    Thanks for finally talking about >Can Handheld XRF Analyze Metal Coating Thickness?
    <Loved it!
    Reply
  4. Logapasana says
    March 2, 2020 at 4:54 am
    Can XRF analyzers can be used to measure or detect the chromate coating on aluminium parts?
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Coating Analysis with XRF
XRF in the Aerospace Industry: Applications in Coating Analysis
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook